Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54619743-54619974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT11
Entrez Gene ID
26027
Gene Name
acyl-CoA thioesterase 11
Gene Aliases
BFIT, STARD14, THEA, THEM1
UniGene
Hs.745173
Gene Chromosome Location
Chr.1: 54548134-54634744 on Build GRCh38
Transcripts
NM_015547.3 NM_176782.2 XM_006710599.3

Gene Symbol
FAM151A
Entrez Gene ID
338094
Gene Name
family with sequence similarity 151 member A
Gene Aliases
C1orf179
UniGene
Hs.234786
Gene Chromosome Location
Chr.1: 54609177-54623527 on Build GRCh38
Transcripts
NM_015547.3 NM_176782.2 XM_006710599.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4166746 c.194G>A p.R65Q Chr.1: 54619932-54619932 on Build GRCh38
5585151 c.171G>A p.L57L Chr.1: 54619955-54619955 on Build GRCh38
1343512 c.222C>T p.H74H Chr.1: 54619904-54619904 on Build GRCh38
6389758 c.227C>A p.A76D Chr.1: 54619899-54619899 on Build GRCh38
1343511 c.226G>A p.A76T Chr.1: 54619900-54619900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144034953 Chr.1: 54619833-54619833 on Build GRCh38
rs768990252 Chr.1: 54619914-54619914 on Build GRCh38
rs763257599 Chr.1: 54619830-54619830 on Build GRCh38
rs745746159 Chr.1: 54619903-54619903 on Build GRCh38
rs147294199 Chr.1: 54619932-54619932 on Build GRCh38
rs770436039 Chr.1: 54619897-54619897 on Build GRCh38
rs114883650 Chr.1: 54619889-54619889 on Build GRCh38
rs963403307 Chr.1: 54619924-54619924 on Build GRCh38
rs200053764 Chr.1: 54619936-54619936 on Build GRCh38
rs779176910 Chr.1: 54619943-54619943 on Build GRCh38
rs781675609 Chr.1: 54619835-54619835 on Build GRCh38
rs747952740 Chr.1: 54619840-54619853 on Build GRCh38
rs954955974 Chr.1: 54619895-54619895 on Build GRCh38
rs776582290 Chr.1: 54619825-54619825 on Build GRCh38
rs11206396 Chr.1: 54619826-54619826 on Build GRCh38
rs886704081 Chr.1: 54619804-54619804 on Build GRCh38
rs115445569 Chr.1: 54619935-54619935 on Build GRCh38
rs17856620 Chr.1: 54619867-54619867 on Build GRCh38
rs1002532343 Chr.1: 54619769-54619769 on Build GRCh38
rs560471881 Chr.1: 54619829-54619829 on Build GRCh38
rs775423980 Chr.1: 54619907-54619907 on Build GRCh38
rs764448487 Chr.1: 54619831-54619831 on Build GRCh38
rs1035726861 Chr.1: 54619777-54619777 on Build GRCh38
rs111721175 Chr.1: 54619871-54619871 on Build GRCh38
rs780576125 Chr.1: 54619900-54619900 on Build GRCh38
rs770752692 Chr.1: 54619819-54619819 on Build GRCh38
rs761940643 Chr.1: 54619915-54619915 on Build GRCh38
rs769912646 Chr.1: 54619904-54619904 on Build GRCh38
rs746664479 Chr.1: 54619890-54619890 on Build GRCh38
rs762887417 Chr.1: 54619908-54619908 on Build GRCh38
rs143150804 Chr.1: 54619933-54619933 on Build GRCh38
rs200790200 Chr.1: 54619864-54619864 on Build GRCh38
rs377683365 Chr.1: 54619852-54619852 on Build GRCh38
rs1005518429 Chr.1: 54619808-54619808 on Build GRCh38
rs149766098 Chr.1: 54619875-54619875 on Build GRCh38
rs756626093 Chr.1: 54619868-54619868 on Build GRCh38
rs746571417 Chr.1: 54619817-54619817 on Build GRCh38
rs371222430 Chr.1: 54619912-54619912 on Build GRCh38

View Full Product Details