Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153976838-153977099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JTB
Entrez Gene ID
10899
Gene Name
jumping translocation breakpoint
Gene Aliases
HJTB, HSPC222, PAR, hJT
UniGene
Hs.6396
Gene Chromosome Location
Chr.1: 153974269-153977975 on Build GRCh38
Transcripts
NM_006694.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527791 c.97C>G p.P33A Chr.1: 153977000-153977000 on Build GRCh38
675201 c.109G>C p.E37Q Chr.1: 153976988-153976988 on Build GRCh38
896481 c.92A>G p.E31G Chr.1: 153977005-153977005 on Build GRCh38
6058039 c.101T>A p.V34E Chr.1: 153976996-153976996 on Build GRCh38
6058040 c.102G>T p.V34V Chr.1: 153976995-153976995 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765157547 Chr.1: 153976944-153976944 on Build GRCh38
rs758001896 Chr.1: 153976949-153976949 on Build GRCh38
rs973562913 Chr.1: 153976921-153976921 on Build GRCh38
rs767485673 Chr.1: 153976857-153976857 on Build GRCh38
rs541400847 Chr.1: 153976912-153976912 on Build GRCh38
rs778131192 Chr.1: 153976976-153976976 on Build GRCh38
rs762749789 Chr.1: 153977026-153977027 on Build GRCh38
rs746057733 Chr.1: 153977000-153977000 on Build GRCh38
rs545656642 Chr.1: 153977070-153977070 on Build GRCh38
rs773146257 Chr.1: 153977030-153977030 on Build GRCh38
rs762872058 Chr.1: 153977046-153977046 on Build GRCh38
rs533464156 Chr.1: 153977067-153977067 on Build GRCh38
rs772734018 Chr.1: 153977020-153977020 on Build GRCh38
rs370055492 Chr.1: 153977009-153977009 on Build GRCh38
rs370802455 Chr.1: 153976952-153976952 on Build GRCh38
rs939870382 Chr.1: 153976875-153976875 on Build GRCh38
rs563830702 Chr.1: 153977077-153977077 on Build GRCh38
rs769102728 Chr.1: 153976961-153976964 on Build GRCh38
rs982375623 Chr.1: 153976861-153976861 on Build GRCh38
rs920754162 Chr.1: 153976923-153976923 on Build GRCh38
rs368680069 Chr.1: 153976986-153976986 on Build GRCh38
rs748691914 Chr.1: 153976961-153976961 on Build GRCh38
rs766073991 Chr.1: 153977054-153977054 on Build GRCh38
rs756157243 Chr.1: 153977062-153977062 on Build GRCh38
rs374447826 Chr.1: 153976956-153976956 on Build GRCh38
rs765990129 Chr.1: 153976950-153976950 on Build GRCh38
rs750195253 Chr.1: 153976946-153976946 on Build GRCh38
rs187859627 Chr.1: 153977060-153977060 on Build GRCh38
rs555366415 Chr.1: 153976859-153976859 on Build GRCh38
rs750472920 Chr.1: 153976919-153976919 on Build GRCh38
rs756570694 Chr.1: 153976964-153976964 on Build GRCh38
rs765208802 Chr.1: 153977029-153977029 on Build GRCh38
rs774697005 Chr.1: 153976997-153976997 on Build GRCh38
rs751271949 Chr.1: 153977057-153977057 on Build GRCh38
rs367803984 Chr.1: 153977018-153977018 on Build GRCh38
rs754574305 Chr.1: 153976953-153976953 on Build GRCh38
rs931116533 Chr.1: 153976951-153976951 on Build GRCh38
rs749591763 Chr.1: 153976983-153976983 on Build GRCh38
rs780249751 Chr.1: 153977072-153977072 on Build GRCh38
rs776761848 Chr.1: 153976930-153976930 on Build GRCh38
rs747312475 Chr.1: 153976936-153976936 on Build GRCh38
rs775673480 Chr.1: 153977017-153977017 on Build GRCh38
rs372071467 Chr.1: 153977059-153977059 on Build GRCh38
rs182139153 Chr.1: 153976895-153976895 on Build GRCh38

View Full Product Details