Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 34945498-34945759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L1
Entrez Gene ID
23333
Gene Name
dpy-19 like 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.408623
Gene Chromosome Location
Chr.7: 34928881-35038041 on Build GRCh38
Transcripts
NM_015283.1 XM_011515245.2 XM_011515246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260573 c.1311A>T p.Q437H Chr.7: 34945681-34945681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758390810 Chr.7: 34945622-34945622 on Build GRCh38
rs367674071 Chr.7: 34945688-34945688 on Build GRCh38
rs765687017 Chr.7: 34945724-34945724 on Build GRCh38
rs762705640 Chr.7: 34945670-34945670 on Build GRCh38
rs924761077 Chr.7: 34945545-34945545 on Build GRCh38
rs746948104 Chr.7: 34945632-34945632 on Build GRCh38
rs201411102 Chr.7: 34945630-34945631 on Build GRCh38
rs756045845 Chr.7: 34945633-34945633 on Build GRCh38
rs774063938 Chr.7: 34945676-34945676 on Build GRCh38
rs541557520 Chr.7: 34945614-34945614 on Build GRCh38
rs758689468 Chr.7: 34945732-34945732 on Build GRCh38
rs761207329 Chr.7: 34945681-34945681 on Build GRCh38
rs758635721 Chr.7: 34945728-34945729 on Build GRCh38
rs370350873 Chr.7: 34945702-34945702 on Build GRCh38
rs745677695 Chr.7: 34945642-34945642 on Build GRCh38
rs777491384 Chr.7: 34945644-34945644 on Build GRCh38
rs556465396 Chr.7: 34945528-34945528 on Build GRCh38
rs561925362 Chr.7: 34945593-34945593 on Build GRCh38
rs993478984 Chr.7: 34945553-34945553 on Build GRCh38
rs374543033 Chr.7: 34945713-34945713 on Build GRCh38
rs1637667 Chr.7: 34945555-34945555 on Build GRCh38
rs114590002 Chr.7: 34945533-34945533 on Build GRCh38
rs774983562 Chr.7: 34945655-34945655 on Build GRCh38
rs879408904 Chr.7: 34945550-34945550 on Build GRCh38
rs564125025 Chr.7: 34945643-34945643 on Build GRCh38
rs752834671 Chr.7: 34945731-34945731 on Build GRCh38
rs777873004 Chr.7: 34945628-34945628 on Build GRCh38
rs527782689 Chr.7: 34945613-34945613 on Build GRCh38
rs780895935 Chr.7: 34945640-34945640 on Build GRCh38
rs768276181 Chr.7: 34945672-34945672 on Build GRCh38

View Full Product Details