Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31333884-31334127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1
Entrez Gene ID
1828
Gene Name
desmoglein 1
Gene Aliases
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1
UniGene
Hs.2633
Gene Chromosome Location
Chr.18: 31318089-31359246 on Build GRCh38
Transcripts
NM_001942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7001586 c.848_849ins16 p.N284fs*3 Chr.18: 31334045-31334046 on Build GRCh38
4634309 c.823A>G p.T275A Chr.18: 31334020-31334020 on Build GRCh38
1716198 c.856A>T p.N286Y Chr.18: 31334053-31334053 on Build GRCh38
5580918 c.924C>T p.F308F Chr.18: 31334121-31334121 on Build GRCh38
3525020 c.865G>A p.E289K Chr.18: 31334062-31334062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950332928 Chr.18: 31334026-31334026 on Build GRCh38
rs796755413 Chr.18: 31333974-31333975 on Build GRCh38
rs150445875 Chr.18: 31334058-31334058 on Build GRCh38
rs752163646 Chr.18: 31334094-31334094 on Build GRCh38
rs138330806 Chr.18: 31334059-31334059 on Build GRCh38
rs377729186 Chr.18: 31334062-31334062 on Build GRCh38
rs761869585 Chr.18: 31334055-31334055 on Build GRCh38
rs113192934 Chr.18: 31333974-31333975 on Build GRCh38
rs978406870 Chr.18: 31333918-31333918 on Build GRCh38
rs201123667 Chr.18: 31334014-31334014 on Build GRCh38
rs369742369 Chr.18: 31333980-31333980 on Build GRCh38
rs778232432 Chr.18: 31334006-31334006 on Build GRCh38
rs776654926 Chr.18: 31334021-31334021 on Build GRCh38
rs556477975 Chr.18: 31333916-31333916 on Build GRCh38
rs145525233 Chr.18: 31333942-31333942 on Build GRCh38
rs917456060 Chr.18: 31334024-31334024 on Build GRCh38
rs915967745 Chr.18: 31333951-31333951 on Build GRCh38
rs754601260 Chr.18: 31334086-31334086 on Build GRCh38
rs760708163 Chr.18: 31333978-31333979 on Build GRCh38
rs775677745 Chr.18: 31334023-31334023 on Build GRCh38
rs1017221849 Chr.18: 31333913-31333913 on Build GRCh38
rs5823800 Chr.18: 31333967-31333968 on Build GRCh38
rs149633304 Chr.18: 31334060-31334060 on Build GRCh38
rs778380651 Chr.18: 31333971-31333971 on Build GRCh38
rs753545622 Chr.18: 31334075-31334075 on Build GRCh38
rs887111701 Chr.18: 31334012-31334012 on Build GRCh38
rs199772847 Chr.18: 31334065-31334065 on Build GRCh38
rs778436186 Chr.18: 31334090-31334090 on Build GRCh38
rs760320030 Chr.18: 31334061-31334061 on Build GRCh38
rs771202624 Chr.18: 31334019-31334019 on Build GRCh38
rs202206724 Chr.18: 31333972-31333972 on Build GRCh38

View Full Product Details