Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82061046-82061293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4070818 c.819C>T p.H273H Chr.17: 82061232-82061232 on Build GRCh38
4508921 c.791C>A p.P264H Chr.17: 82061260-82061260 on Build GRCh38
6056728 c.769C>A p.L257M Chr.17: 82061282-82061282 on Build GRCh38
4508474 c.777C>T p.I259I Chr.17: 82061274-82061274 on Build GRCh38
4985757 c.812G>C p.R271P Chr.17: 82061239-82061239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527980841 Chr.17: 82061188-82061188 on Build GRCh38
rs752083808 Chr.17: 82061180-82061180 on Build GRCh38
rs768911603 Chr.17: 82061206-82061207 on Build GRCh38
rs369444572 Chr.17: 82061262-82061262 on Build GRCh38
rs778481501 Chr.17: 82061209-82061209 on Build GRCh38
rs775756471 Chr.17: 82061172-82061172 on Build GRCh38
rs777649647 Chr.17: 82061213-82061213 on Build GRCh38
rs368812276 Chr.17: 82061170-82061170 on Build GRCh38
rs748078918 Chr.17: 82061251-82061251 on Build GRCh38
rs541939857 Chr.17: 82061103-82061103 on Build GRCh38
rs1049839424 Chr.17: 82061154-82061154 on Build GRCh38
rs753416829 Chr.17: 82061198-82061198 on Build GRCh38
rs775607556 Chr.17: 82061253-82061253 on Build GRCh38
rs529008524 Chr.17: 82061252-82061252 on Build GRCh38
rs922616837 Chr.17: 82061142-82061142 on Build GRCh38
rs768075209 Chr.17: 82061270-82061270 on Build GRCh38
rs781397100 Chr.17: 82061251-82061252 on Build GRCh38
rs1049142440 Chr.17: 82061065-82061065 on Build GRCh38
rs762164701 Chr.17: 82061269-82061269 on Build GRCh38
rs574621918 Chr.17: 82061081-82061081 on Build GRCh38
rs867313731 Chr.17: 82061096-82061096 on Build GRCh38
rs1037230683 Chr.17: 82061137-82061137 on Build GRCh38
rs375227321 Chr.17: 82061244-82061244 on Build GRCh38
rs556373339 Chr.17: 82061074-82061074 on Build GRCh38
rs901631135 Chr.17: 82061120-82061120 on Build GRCh38
rs893631956 Chr.17: 82061199-82061199 on Build GRCh38
rs776784588 Chr.17: 82061162-82061162 on Build GRCh38
rs200310914 Chr.17: 82061190-82061190 on Build GRCh38
rs769884511 Chr.17: 82061251-82061251 on Build GRCh38
rs889924009 Chr.17: 82061066-82061066 on Build GRCh38
rs200452679 Chr.17: 82061181-82061181 on Build GRCh38
rs1053643088 Chr.17: 82061197-82061197 on Build GRCh38
rs754501147 Chr.17: 82061208-82061208 on Build GRCh38
rs997164997 Chr.17: 82061140-82061140 on Build GRCh38
rs199812921 Chr.17: 82061239-82061239 on Build GRCh38
rs758147265 Chr.17: 82061211-82061211 on Build GRCh38
rs765626186 Chr.17: 82061152-82061152 on Build GRCh38
rs759683343 Chr.17: 82061163-82061163 on Build GRCh38
rs763269004 Chr.17: 82061179-82061179 on Build GRCh38
rs762378509 Chr.17: 82061085-82061085 on Build GRCh38
rs140560459 Chr.17: 82061220-82061220 on Build GRCh38
rs940299533 Chr.17: 82061135-82061135 on Build GRCh38
rs769022278 Chr.17: 82061256-82061256 on Build GRCh38
rs35439399 Chr.17: 82061071-82061071 on Build GRCh38
rs372820618 Chr.17: 82061240-82061240 on Build GRCh38
rs36067039 Chr.17: 82061236-82061237 on Build GRCh38

View Full Product Details