Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29575110-29575337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT1
Entrez Gene ID
28964
Gene Name
GIT ArfGAP 1
Gene Aliases
-
UniGene
Hs.514051
Gene Chromosome Location
Chr.17: 29573469-29589592 on Build GRCh38
Transcripts
NM_001085454.1 NM_014030.3 XM_011524684.1 XM_011524685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362058 c.2008C>A p.R670R Chr.17: 29575316-29575316 on Build GRCh38
977191 c.2002C>A p.L668M Chr.17: 29575322-29575322 on Build GRCh38
6671074 c.1980G>A p.L660L Chr.17: 29575317-29575317 on Build GRCh38
4598506 c.2008C>T p.R670W Chr.17: 29575316-29575316 on Build GRCh38
5740931 c.2012C>T p.A671V Chr.17: 29575312-29575312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545018364 Chr.17: 29575232-29575232 on Build GRCh38
rs1048582563 Chr.17: 29575265-29575265 on Build GRCh38
rs1016095545 Chr.17: 29575280-29575280 on Build GRCh38
rs377279123 Chr.17: 29575148-29575148 on Build GRCh38
rs373722852 Chr.17: 29575176-29575176 on Build GRCh38
rs766906155 Chr.17: 29575290-29575290 on Build GRCh38
rs762565788 Chr.17: 29575299-29575299 on Build GRCh38
rs925891052 Chr.17: 29575315-29575315 on Build GRCh38
rs768890171 Chr.17: 29575269-29575269 on Build GRCh38
rs761903268 Chr.17: 29575272-29575272 on Build GRCh38
rs372900000 Chr.17: 29575149-29575149 on Build GRCh38
rs1046066975 Chr.17: 29575251-29575251 on Build GRCh38
rs757168583 Chr.17: 29575191-29575191 on Build GRCh38
rs749537581 Chr.17: 29575267-29575267 on Build GRCh38
rs377389432 Chr.17: 29575282-29575282 on Build GRCh38
rs965496338 Chr.17: 29575227-29575227 on Build GRCh38
rs200748938 Chr.17: 29575183-29575183 on Build GRCh38
rs774342882 Chr.17: 29575270-29575270 on Build GRCh38
rs751066009 Chr.17: 29575308-29575308 on Build GRCh38
rs145908069 Chr.17: 29575247-29575247 on Build GRCh38
rs769522976 Chr.17: 29575264-29575264 on Build GRCh38
rs905460658 Chr.17: 29575258-29575258 on Build GRCh38
rs575569631 Chr.17: 29575248-29575248 on Build GRCh38
rs542847680 Chr.17: 29575302-29575302 on Build GRCh38
rs369310943 Chr.17: 29575157-29575157 on Build GRCh38
rs145991218 Chr.17: 29575138-29575138 on Build GRCh38
rs552780242 Chr.17: 29575184-29575184 on Build GRCh38
rs546843 Chr.17: 29575187-29575187 on Build GRCh38
rs768114997 Chr.17: 29575158-29575158 on Build GRCh38
rs754202696 Chr.17: 29575293-29575293 on Build GRCh38
rs756503801 Chr.17: 29575173-29575173 on Build GRCh38
rs745617945 Chr.17: 29575139-29575139 on Build GRCh38
rs775624860 Chr.17: 29575147-29575147 on Build GRCh38
rs773784492 Chr.17: 29575275-29575275 on Build GRCh38
rs181497741 Chr.17: 29575155-29575155 on Build GRCh38
rs555497918 Chr.17: 29575146-29575146 on Build GRCh38
rs746639574 Chr.17: 29575230-29575230 on Build GRCh38
rs746510074 Chr.17: 29575190-29575190 on Build GRCh38
rs577598394 Chr.17: 29575221-29575221 on Build GRCh38
rs772732892 Chr.17: 29575274-29575274 on Build GRCh38
rs745668975 Chr.17: 29575262-29575262 on Build GRCh38
rs750836667 Chr.17: 29575172-29575172 on Build GRCh38
rs369478533 Chr.17: 29575257-29575257 on Build GRCh38

View Full Product Details