Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 91079313-91079586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
UniGene
Hs.258576
Gene Chromosome Location
Chr.7: 90595994-91210590 on Build GRCh38
Transcripts
NM_001287135.1 NM_001287136.1 NM_001287137.1 NM_012395.3 XM_005250436.3 XM_005250438.3 XM_005250439.2 XM_011516306.2 XM_017012320.1 XM_017012321.1 XM_017012322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7117161 c.1085G>C p.R362T Chr.7: 91079465-91079465 on Build GRCh38
6625392 c.1100+1G>A p.? Chr.7: 91079481-91079481 on Build GRCh38
1247925 c.1100A>C p.K367T Chr.7: 91079480-91079480 on Build GRCh38
6625393 c.1154+1G>A p.? Chr.7: 91079481-91079481 on Build GRCh38
7117162 c.1139G>C p.R380T Chr.7: 91079465-91079465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544929388 Chr.7: 91079549-91079549 on Build GRCh38
rs781363070 Chr.7: 91079432-91079432 on Build GRCh38
rs752813423 Chr.7: 91079532-91079532 on Build GRCh38
rs145058405 Chr.7: 91079462-91079462 on Build GRCh38
rs762435383 Chr.7: 91079395-91079395 on Build GRCh38
rs112473985 Chr.7: 91079487-91079487 on Build GRCh38
rs918764879 Chr.7: 91079389-91079389 on Build GRCh38
rs763491903 Chr.7: 91079511-91079511 on Build GRCh38
rs1047236355 Chr.7: 91079384-91079384 on Build GRCh38
rs763594213 Chr.7: 91079528-91079534 on Build GRCh38
rs776256901 Chr.7: 91079475-91079475 on Build GRCh38
rs770487698 Chr.7: 91079494-91079494 on Build GRCh38
rs748125989 Chr.7: 91079434-91079434 on Build GRCh38
rs929050397 Chr.7: 91079347-91079347 on Build GRCh38
rs191632826 Chr.7: 91079369-91079369 on Build GRCh38
rs573708597 Chr.7: 91079385-91079385 on Build GRCh38
rs199778971 Chr.7: 91079519-91079519 on Build GRCh38
rs755087534 Chr.7: 91079425-91079425 on Build GRCh38
rs773839618 Chr.7: 91079495-91079495 on Build GRCh38
rs575758767 Chr.7: 91079546-91079546 on Build GRCh38
rs34419285 Chr.7: 91079461-91079461 on Build GRCh38
rs552949476 Chr.7: 91079442-91079442 on Build GRCh38
rs767612297 Chr.7: 91079531-91079531 on Build GRCh38
rs751816082 Chr.7: 91079421-91079421 on Build GRCh38
rs771808000 Chr.7: 91079361-91079361 on Build GRCh38
rs908721787 Chr.7: 91079409-91079409 on Build GRCh38
rs759824402 Chr.7: 91079525-91079525 on Build GRCh38
rs972988251 Chr.7: 91079358-91079358 on Build GRCh38
rs1053798846 Chr.7: 91079548-91079548 on Build GRCh38
rs377019818 Chr.7: 91079403-91079403 on Build GRCh38
rs758828432 Chr.7: 91079412-91079412 on Build GRCh38
rs942444813 Chr.7: 91079467-91079467 on Build GRCh38
rs762590788 Chr.7: 91079507-91079523 on Build GRCh38
rs766767747 Chr.7: 91079515-91079515 on Build GRCh38
rs774836265 Chr.7: 91079381-91079381 on Build GRCh38
rs778818423 Chr.7: 91079435-91079435 on Build GRCh38
rs941632722 Chr.7: 91079342-91079342 on Build GRCh38
rs766563091 Chr.7: 91079418-91079418 on Build GRCh38
rs573288226 Chr.7: 91079449-91079449 on Build GRCh38
rs765819597 Chr.7: 91079397-91079397 on Build GRCh38
rs768148638 Chr.7: 91079473-91079473 on Build GRCh38
rs536969461 Chr.7: 91079336-91079336 on Build GRCh38
rs542569374 Chr.7: 91079394-91079394 on Build GRCh38
rs775081722 Chr.7: 91079459-91079459 on Build GRCh38

View Full Product Details