Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105030287-105030558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP1
Entrez Gene ID
834
Gene Name
caspase 1
Gene Aliases
ICE, IL1BC, P45
UniGene
Hs.2490
Gene Chromosome Location
Chr.11: 105025508-105035591 on Build GRCh38
Transcripts
NM_001223.4 NM_001257118.2 NM_001257119.2 NM_033292.3 NM_033293.3 NM_033294.3 NM_033295.3 XM_017018393.1 XM_017018394.1 XM_017018395.1 XM_017018396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2109182 c.507C>T p.C169C Chr.11: 105030450-105030450 on Build GRCh38
6222049 c.613_614insA p.N205fs*24 Chr.11: 105030343-105030344 on Build GRCh38
1199639 c.607A>G p.K203E Chr.11: 105030350-105030350 on Build GRCh38
2109181 c.609A>G p.K203K Chr.11: 105030348-105030348 on Build GRCh38
5808518 c.487C>G p.R163G Chr.11: 105030470-105030470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763427774 Chr.11: 105030461-105030461 on Build GRCh38
rs765739042 Chr.11: 105030349-105030349 on Build GRCh38
rs376822417 Chr.11: 105030341-105030341 on Build GRCh38
rs754542264 Chr.11: 105030354-105030354 on Build GRCh38
rs771578706 Chr.11: 105030335-105030336 on Build GRCh38
rs780648592 Chr.11: 105030359-105030359 on Build GRCh38
rs746543598 Chr.11: 105030343-105030343 on Build GRCh38
rs755812705 Chr.11: 105030383-105030383 on Build GRCh38
rs897409593 Chr.11: 105030459-105030459 on Build GRCh38
rs371409713 Chr.11: 105030431-105030431 on Build GRCh38
rs759699472 Chr.11: 105030326-105030326 on Build GRCh38
rs143461834 Chr.11: 105030335-105030335 on Build GRCh38
rs759796120 Chr.11: 105030416-105030416 on Build GRCh38
rs762044422 Chr.11: 105030345-105030345 on Build GRCh38
rs780307078 Chr.11: 105030498-105030499 on Build GRCh38
rs199667142 Chr.11: 105030330-105030330 on Build GRCh38
rs371404424 Chr.11: 105030379-105030379 on Build GRCh38
rs752394637 Chr.11: 105030360-105030360 on Build GRCh38
rs373584770 Chr.11: 105030337-105030337 on Build GRCh38
rs777246560 Chr.11: 105030386-105030386 on Build GRCh38
rs757139801 Chr.11: 105030485-105030485 on Build GRCh38
rs753417252 Chr.11: 105030478-105030478 on Build GRCh38
rs994248249 Chr.11: 105030362-105030362 on Build GRCh38
rs562777550 Chr.11: 105030491-105030491 on Build GRCh38
rs765575404 Chr.11: 105030451-105030451 on Build GRCh38
rs752283506 Chr.11: 105030470-105030470 on Build GRCh38
rs770928654 Chr.11: 105030312-105030312 on Build GRCh38
rs766822009 Chr.11: 105030469-105030469 on Build GRCh38
rs760923024 Chr.11: 105030334-105030334 on Build GRCh38
rs140337178 Chr.11: 105030401-105030401 on Build GRCh38
rs557462109 Chr.11: 105030316-105030316 on Build GRCh38
rs778821962 Chr.11: 105030490-105030490 on Build GRCh38
rs768253520 Chr.11: 105030342-105030343 on Build GRCh38
rs61755276 Chr.11: 105030331-105030331 on Build GRCh38
rs779588701 Chr.11: 105030342-105030342 on Build GRCh38
rs776651091 Chr.11: 105030444-105030444 on Build GRCh38
rs137959643 Chr.11: 105030477-105030477 on Build GRCh38
rs377331184 Chr.11: 105030476-105030476 on Build GRCh38
rs376117763 Chr.11: 105030455-105030455 on Build GRCh38
rs760626645 Chr.11: 105030442-105030442 on Build GRCh38
rs768953462 Chr.11: 105030443-105030443 on Build GRCh38
rs762252349 Chr.11: 105030447-105030447 on Build GRCh38
rs780103519 Chr.11: 105030506-105030506 on Build GRCh38
rs750873942 Chr.11: 105030353-105030353 on Build GRCh38
rs145465840 Chr.11: 105030427-105030427 on Build GRCh38
rs750966281 Chr.11: 105030458-105030458 on Build GRCh38
rs531803659 Chr.11: 105030498-105030498 on Build GRCh38
rs749109400 Chr.11: 105030400-105030400 on Build GRCh38
rs994214999 Chr.11: 105030508-105030508 on Build GRCh38

View Full Product Details