Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233158344-233158605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
UniGene
Hs.262886
Gene Chromosome Location
Chr.2: 233059967-233207903 on Build GRCh38
Transcripts
NM_001017915.2 NM_005541.4 XM_017004004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5532738 c.1109G>A p.R370Q Chr.2: 233158391-233158391 on Build GRCh38
5396494 c.1114G>A p.E372K Chr.2: 233158396-233158396 on Build GRCh38
3962543 c.1135A>G p.K379E Chr.2: 233158417-233158417 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs36181983 Chr.2: 233158580-233158580 on Build GRCh38
rs148118779 Chr.2: 233158371-233158371 on Build GRCh38
rs919389629 Chr.2: 233158566-233158566 on Build GRCh38
rs36153082 Chr.2: 233158577-233158577 on Build GRCh38
rs150623170 Chr.2: 233158489-233158489 on Build GRCh38
rs958523454 Chr.2: 233158507-233158507 on Build GRCh38
rs567316178 Chr.2: 233158415-233158415 on Build GRCh38
rs546332120 Chr.2: 233158383-233158383 on Build GRCh38
rs769652125 Chr.2: 233158430-233158430 on Build GRCh38
rs1011516902 Chr.2: 233158464-233158464 on Build GRCh38
rs767727828 Chr.2: 233158462-233158462 on Build GRCh38
rs762086030 Chr.2: 233158446-233158446 on Build GRCh38
rs558608037 Chr.2: 233158519-233158519 on Build GRCh38
rs373012211 Chr.2: 233158490-233158490 on Build GRCh38
rs967217621 Chr.2: 233158486-233158486 on Build GRCh38
rs893028977 Chr.2: 233158448-233158448 on Build GRCh38
rs537736321 Chr.2: 233158560-233158560 on Build GRCh38
rs1036871661 Chr.2: 233158496-233158496 on Build GRCh38
rs1020107279 Chr.2: 233158474-233158474 on Build GRCh38
rs36165346 Chr.2: 233158443-233158443 on Build GRCh38
rs1051887386 Chr.2: 233158515-233158521 on Build GRCh38
rs933820191 Chr.2: 233158513-233158514 on Build GRCh38
rs963940181 Chr.2: 233158546-233158546 on Build GRCh38
rs1031919490 Chr.2: 233158581-233158581 on Build GRCh38
rs538882093 Chr.2: 233158534-233158534 on Build GRCh38
rs941289982 Chr.2: 233158492-233158492 on Build GRCh38
rs972299947 Chr.2: 233158557-233158557 on Build GRCh38
rs138944017 Chr.2: 233158545-233158545 on Build GRCh38
rs1035444375 Chr.2: 233158499-233158499 on Build GRCh38
rs1007612406 Chr.2: 233158542-233158542 on Build GRCh38

View Full Product Details