Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12924634-12924859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSA
Entrez Gene ID
2193
Gene Name
phenylalanyl-tRNA synthetase alpha subunit
Gene Aliases
CML33, FARSL, FARSLA, FRSA, PheHA
UniGene
Hs.23111
Gene Chromosome Location
Chr.19: 12922470-12933744 on Build GRCh38
Transcripts
NM_004461.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5818905 c.1128G>T p.V376V Chr.19: 12924706-12924706 on Build GRCh38
1206539 c.1174C>T p.R392W Chr.19: 12924660-12924660 on Build GRCh38
7414103 c.1072C>T p.R358W Chr.19: 12924762-12924762 on Build GRCh38
6149579 c.1131G>A p.V377V Chr.19: 12924703-12924703 on Build GRCh38
4405572 c.1106A>C p.E369A Chr.19: 12924728-12924728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754958871 Chr.19: 12924827-12924827 on Build GRCh38
rs140193183 Chr.19: 12924708-12924708 on Build GRCh38
rs745947510 Chr.19: 12924744-12924744 on Build GRCh38
rs769334009 Chr.19: 12924770-12924770 on Build GRCh38
rs375677041 Chr.19: 12924666-12924666 on Build GRCh38
rs768613348 Chr.19: 12924736-12924736 on Build GRCh38
rs764423533 Chr.19: 12924726-12924726 on Build GRCh38
rs956549902 Chr.19: 12924682-12924682 on Build GRCh38
rs750617542 Chr.19: 12924822-12924822 on Build GRCh38
rs764856282 Chr.19: 12924701-12924701 on Build GRCh38
rs756425459 Chr.19: 12924655-12924655 on Build GRCh38
rs779126069 Chr.19: 12924740-12924740 on Build GRCh38
rs768617085 Chr.19: 12924750-12924750 on Build GRCh38
rs576348484 Chr.19: 12924684-12924684 on Build GRCh38
rs139805483 Chr.19: 12924794-12924794 on Build GRCh38
rs778006163 Chr.19: 12924659-12924659 on Build GRCh38
rs747899400 Chr.19: 12924769-12924769 on Build GRCh38
rs148613387 Chr.19: 12924700-12924700 on Build GRCh38
rs762404457 Chr.19: 12924774-12924774 on Build GRCh38
rs200214203 Chr.19: 12924837-12924837 on Build GRCh38
rs147769605 Chr.19: 12924745-12924745 on Build GRCh38
rs762456288 Chr.19: 12924706-12924706 on Build GRCh38
rs745886681 Chr.19: 12924667-12924667 on Build GRCh38
rs765847659 Chr.19: 12924775-12924775 on Build GRCh38
rs560812068 Chr.19: 12924823-12924823 on Build GRCh38
rs754160224 Chr.19: 12924800-12924800 on Build GRCh38
rs530112428 Chr.19: 12924793-12924793 on Build GRCh38
rs368255552 Chr.19: 12924816-12924816 on Build GRCh38
rs143923872 Chr.19: 12924695-12924695 on Build GRCh38
rs8107173 Chr.19: 12924824-12924824 on Build GRCh38
rs771092341 Chr.19: 12924663-12924663 on Build GRCh38
rs746912275 Chr.19: 12924748-12924748 on Build GRCh38
rs537993454 Chr.19: 12924835-12924835 on Build GRCh38
rs772874371 Chr.19: 12924771-12924771 on Build GRCh38
rs921955922 Chr.19: 12924731-12924731 on Build GRCh38
rs374755594 Chr.19: 12924739-12924739 on Build GRCh38
rs756513794 Chr.19: 12924709-12924709 on Build GRCh38
rs776029885 Chr.19: 12924778-12924778 on Build GRCh38
rs371247417 Chr.19: 12924761-12924761 on Build GRCh38
rs754147656 Chr.19: 12924727-12924727 on Build GRCh38
rs749767689 Chr.19: 12924687-12924688 on Build GRCh38
rs113870503 Chr.19: 12924703-12924703 on Build GRCh38
rs759933679 Chr.19: 12924831-12924831 on Build GRCh38
rs201131468 Chr.19: 12924672-12924672 on Build GRCh38
rs757585686 Chr.19: 12924814-12924814 on Build GRCh38
rs749443581 Chr.19: 12924660-12924660 on Build GRCh38
rs771909644 Chr.19: 12924668-12924668 on Build GRCh38
rs746820628 Chr.19: 12924679-12924679 on Build GRCh38
rs776462226 Chr.19: 12924690-12924690 on Build GRCh38

View Full Product Details