Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73500503-73500776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT1
Entrez Gene ID
641371
Gene Name
acyl-CoA thioesterase 1
Gene Aliases
ACH2, CTE-1, LACH2
UniGene
Hs.446685
Gene Chromosome Location
Chr.14: 73493021-73543799 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
XM_017021590.1 XM_017021591.1 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4052280 c.2005G>A p.A669T Chr.14: 73500690-73500690 on Build GRCh38
7134648 c.2126T>G p.L709R Chr.14: 73500569-73500569 on Build GRCh38
3936603 c.2157G>C p.L719L Chr.14: 73500679-73500679 on Build GRCh38
6418127 c.2278G>A p.D760N Chr.14: 73500558-73500558 on Build GRCh38
6386829 c.2041G>A p.A681T Chr.14: 73500654-73500654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778213775 Chr.14: 73500582-73500582 on Build GRCh38
rs201492591 Chr.14: 73500639-73500639 on Build GRCh38
rs927659000 Chr.14: 73500583-73500583 on Build GRCh38
rs990375673 Chr.14: 73500538-73500538 on Build GRCh38
rs765867302 Chr.14: 73500683-73500684 on Build GRCh38
rs200275195 Chr.14: 73500605-73500605 on Build GRCh38
rs997723679 Chr.14: 73500572-73500572 on Build GRCh38
rs752962339 Chr.14: 73500651-73500651 on Build GRCh38
rs201799912 Chr.14: 73500596-73500596 on Build GRCh38
rs754322974 Chr.14: 73500597-73500597 on Build GRCh38
rs757798640 Chr.14: 73500679-73500679 on Build GRCh38
rs141137854 Chr.14: 73500698-73500698 on Build GRCh38
rs1054837312 Chr.14: 73500587-73500587 on Build GRCh38
rs200844564 Chr.14: 73500743-73500743 on Build GRCh38
rs570028550 Chr.14: 73500599-73500599 on Build GRCh38
rs771041907 Chr.14: 73500723-73500723 on Build GRCh38
rs760642546 Chr.14: 73500645-73500645 on Build GRCh38
rs546374990 Chr.14: 73500535-73500535 on Build GRCh38
rs751097810 Chr.14: 73500696-73500697 on Build GRCh38
rs200682028 Chr.14: 73500700-73500700 on Build GRCh38
rs148646899 Chr.14: 73500561-73500561 on Build GRCh38
rs776891385 Chr.14: 73500648-73500648 on Build GRCh38
rs753400406 Chr.14: 73500693-73500693 on Build GRCh38
rs767412277 Chr.14: 73500551-73500551 on Build GRCh38
rs747452559 Chr.14: 73500595-73500595 on Build GRCh38
rs538257935 Chr.14: 73500559-73500559 on Build GRCh38
rs201293726 Chr.14: 73500606-73500606 on Build GRCh38
rs752089743 Chr.14: 73500658-73500658 on Build GRCh38
rs571262582 Chr.14: 73500558-73500558 on Build GRCh38
rs1051154446 Chr.14: 73500598-73500598 on Build GRCh38
rs752940929 Chr.14: 73500720-73500720 on Build GRCh38
rs777673912 Chr.14: 73500702-73500702 on Build GRCh38
rs771832393 Chr.14: 73500610-73500610 on Build GRCh38
rs747777053 Chr.14: 73500699-73500699 on Build GRCh38
rs371496251 Chr.14: 73500560-73500560 on Build GRCh38
rs763234344 Chr.14: 73500654-73500654 on Build GRCh38
rs755395364 Chr.14: 73500608-73500608 on Build GRCh38
rs371493329 Chr.14: 73500650-73500650 on Build GRCh38
rs774036680 Chr.14: 73500603-73500603 on Build GRCh38
rs781645311 Chr.14: 73500683-73500683 on Build GRCh38
rs151153984 Chr.14: 73500655-73500655 on Build GRCh38
rs768238517 Chr.14: 73500600-73500600 on Build GRCh38
rs766598807 Chr.14: 73500646-73500646 on Build GRCh38
rs776696705 Chr.14: 73500724-73500724 on Build GRCh38

View Full Product Details