Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45544767-45545014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6B
Entrez Gene ID
220081
Gene Name
glutamate rich 6B
Gene Aliases
FAM194B
UniGene
Hs.668747
Gene Chromosome Location
Chr.13: 45541203-45615739 on Build GRCh38
Transcripts
NM_182542.2 XM_011534965.2 XM_017020418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1367023 c.1823T>C p.F608S Chr.13: 45544809-45544809 on Build GRCh38
4472353 c.1779C>A p.Y593* Chr.13: 45544853-45544853 on Build GRCh38
5909332 c.1716G>A p.W572* Chr.13: 45544916-45544916 on Build GRCh38
6368633 c.1738G>A p.A580T Chr.13: 45544894-45544894 on Build GRCh38
5493096 c.1693C>T p.R565C Chr.13: 45544939-45544939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049386422 Chr.13: 45544832-45544832 on Build GRCh38
rs376048631 Chr.13: 45544941-45544941 on Build GRCh38
rs886160689 Chr.13: 45544862-45544862 on Build GRCh38
rs867618281 Chr.13: 45544792-45544792 on Build GRCh38
rs142656281 Chr.13: 45544833-45544833 on Build GRCh38
rs931980874 Chr.13: 45544840-45544840 on Build GRCh38
rs112232111 Chr.13: 45544894-45544894 on Build GRCh38
rs1039976572 Chr.13: 45544917-45544917 on Build GRCh38
rs1031185541 Chr.13: 45544985-45544985 on Build GRCh38
rs569722960 Chr.13: 45544950-45544950 on Build GRCh38
rs879164523 Chr.13: 45544947-45544947 on Build GRCh38
rs374526358 Chr.13: 45544902-45544902 on Build GRCh38
rs527763848 Chr.13: 45544887-45544887 on Build GRCh38
rs911980556 Chr.13: 45544883-45544883 on Build GRCh38
rs552252490 Chr.13: 45544968-45544968 on Build GRCh38
rs752935669 Chr.13: 45544908-45544908 on Build GRCh38
rs770926604 Chr.13: 45544842-45544842 on Build GRCh38
rs751568577 Chr.13: 45544956-45544956 on Build GRCh38
rs996821525 Chr.13: 45544928-45544928 on Build GRCh38
rs901555360 Chr.13: 45544919-45544919 on Build GRCh38
rs537055544 Chr.13: 45544986-45544986 on Build GRCh38
rs1051813501 Chr.13: 45544882-45544882 on Build GRCh38
rs41292935 Chr.13: 45544895-45544895 on Build GRCh38
rs780390003 Chr.13: 45544835-45544835 on Build GRCh38
rs7327901 Chr.13: 45544939-45544939 on Build GRCh38
rs560770493 Chr.13: 45544805-45544805 on Build GRCh38
rs930517957 Chr.13: 45544809-45544809 on Build GRCh38
rs927107227 Chr.13: 45544803-45544803 on Build GRCh38
rs758563788 Chr.13: 45544938-45544938 on Build GRCh38
rs371266878 Chr.13: 45544963-45544963 on Build GRCh38

View Full Product Details