Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1496249-1496474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5K
Entrez Gene ID
51763
Gene Name
inositol polyphosphate-5-phosphatase K
Gene Aliases
PPS, SKIP
UniGene
Hs.632238
Gene Chromosome Location
Chr.17: 1494575-1516888 on Build GRCh38
Transcripts
NM_001135642.1 NM_016532.3 NM_130766.2 XM_005256683.2 XM_005256685.1 XM_005256686.1 XM_011523934.1 XM_011523936.2 XM_017024755.1 XM_017024756.1 XM_017024757.1 XM_017024758.1 XM_017024759.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5032398 c.874-6T>C p.? Chr.17: 1496408-1496408 on Build GRCh38
3701154 c.915G>T p.W305C Chr.17: 1496361-1496361 on Build GRCh38
3701153 c.1143G>T p.W381C Chr.17: 1496361-1496361 on Build GRCh38
4608648 c.910G>T p.A304S Chr.17: 1496366-1496366 on Build GRCh38
7324773 c.1185+2T>C p.? Chr.17: 1496317-1496317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765374051 Chr.17: 1496414-1496414 on Build GRCh38
rs144837901 Chr.17: 1496333-1496333 on Build GRCh38
rs147450510 Chr.17: 1496300-1496300 on Build GRCh38
rs761008667 Chr.17: 1496296-1496296 on Build GRCh38
rs757415734 Chr.17: 1496314-1496314 on Build GRCh38
rs969750985 Chr.17: 1496297-1496297 on Build GRCh38
rs777534819 Chr.17: 1496352-1496352 on Build GRCh38
rs866730600 Chr.17: 1496393-1496393 on Build GRCh38
rs114221242 Chr.17: 1496292-1496292 on Build GRCh38
rs928339424 Chr.17: 1496306-1496306 on Build GRCh38
rs767344347 Chr.17: 1496362-1496362 on Build GRCh38
rs1023854665 Chr.17: 1496421-1496421 on Build GRCh38
rs566828608 Chr.17: 1496358-1496358 on Build GRCh38
rs780421732 Chr.17: 1496332-1496332 on Build GRCh38
rs775485102 Chr.17: 1496349-1496349 on Build GRCh38
rs372068677 Chr.17: 1496376-1496376 on Build GRCh38
rs951553447 Chr.17: 1496304-1496304 on Build GRCh38
rs866371136 Chr.17: 1496357-1496357 on Build GRCh38
rs912591630 Chr.17: 1496312-1496312 on Build GRCh38
rs367903842 Chr.17: 1496319-1496319 on Build GRCh38
rs375291449 Chr.17: 1496310-1496310 on Build GRCh38
rs61729422 Chr.17: 1496328-1496328 on Build GRCh38
rs949200099 Chr.17: 1496374-1496374 on Build GRCh38
rs767433598 Chr.17: 1496283-1496283 on Build GRCh38
rs755810408 Chr.17: 1496391-1496391 on Build GRCh38
rs970981429 Chr.17: 1496448-1496448 on Build GRCh38
rs1046140838 Chr.17: 1496375-1496375 on Build GRCh38
rs139890859 Chr.17: 1496355-1496355 on Build GRCh38
rs760595645 Chr.17: 1496381-1496381 on Build GRCh38
rs991508158 Chr.17: 1496271-1496271 on Build GRCh38
rs867507549 Chr.17: 1496356-1496356 on Build GRCh38
rs145752810 Chr.17: 1496334-1496334 on Build GRCh38
rs753840406 Chr.17: 1496301-1496301 on Build GRCh38
rs148962273 Chr.17: 1496364-1496364 on Build GRCh38
rs558584523 Chr.17: 1496392-1496392 on Build GRCh38
rs1001859918 Chr.17: 1496425-1496425 on Build GRCh38
rs867057266 Chr.17: 1496365-1496365 on Build GRCh38
rs1015859902 Chr.17: 1496426-1496427 on Build GRCh38
rs868335471 Chr.17: 1496327-1496327 on Build GRCh38
rs761735881 Chr.17: 1496411-1496411 on Build GRCh38
rs779287248 Chr.17: 1496315-1496315 on Build GRCh38

View Full Product Details