Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36284370-36284631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 NM_001170700.2 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634576 c.364G>T p.E122* Chr.4: 36284443-36284443 on Build GRCh38
5995634 c.372A>T p.E124D Chr.4: 36284451-36284451 on Build GRCh38
7427768 c.387A>T p.S129S Chr.4: 36284466-36284466 on Build GRCh38
6575693 c.339G>C p.E113D Chr.4: 36284418-36284418 on Build GRCh38
3825781 c.322A>C p.N108H Chr.4: 36284401-36284401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924160976 Chr.4: 36284513-36284513 on Build GRCh38
rs753725419 Chr.4: 36284603-36284603 on Build GRCh38
rs34346982 Chr.4: 36284410-36284411 on Build GRCh38
rs930144559 Chr.4: 36284438-36284438 on Build GRCh38
rs767273674 Chr.4: 36284534-36284534 on Build GRCh38
rs1030877579 Chr.4: 36284578-36284578 on Build GRCh38
rs1042634650 Chr.4: 36284510-36284510 on Build GRCh38
rs922253182 Chr.4: 36284466-36284466 on Build GRCh38
rs765242624 Chr.4: 36284447-36284447 on Build GRCh38
rs976421343 Chr.4: 36284454-36284454 on Build GRCh38
rs766301788 Chr.4: 36284560-36284560 on Build GRCh38
rs73229054 Chr.4: 36284538-36284538 on Build GRCh38
rs377740589 Chr.4: 36284462-36284462 on Build GRCh38
rs956587496 Chr.4: 36284580-36284580 on Build GRCh38
rs896581316 Chr.4: 36284492-36284492 on Build GRCh38
rs73117663 Chr.4: 36284605-36284605 on Build GRCh38
rs763224410 Chr.4: 36284520-36284520 on Build GRCh38
rs868641702 Chr.4: 36284528-36284528 on Build GRCh38
rs184173479 Chr.4: 36284440-36284440 on Build GRCh38
rs1031670278 Chr.4: 36284491-36284491 on Build GRCh38
rs1000778156 Chr.4: 36284573-36284573 on Build GRCh38
rs780831594 Chr.4: 36284545-36284546 on Build GRCh38
rs918804608 Chr.4: 36284406-36284406 on Build GRCh38
rs895240671 Chr.4: 36284590-36284590 on Build GRCh38
rs547413369 Chr.4: 36284411-36284411 on Build GRCh38
rs1054325137 Chr.4: 36284546-36284546 on Build GRCh38
rs755927196 Chr.4: 36284547-36284547 on Build GRCh38
rs1046421888 Chr.4: 36284607-36284607 on Build GRCh38
rs189968146 Chr.4: 36284553-36284553 on Build GRCh38

View Full Product Details