Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73762622-73762869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 XM_017010283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175293 c.895A>C p.M299L Chr.6: 73762780-73762780 on Build GRCh38
4586918 c.857T>C p.I286T Chr.6: 73762742-73762742 on Build GRCh38
3831031 c.917C>T p.S306L Chr.6: 73762802-73762802 on Build GRCh38
6410273 c.866C>A p.S289Y Chr.6: 73762751-73762751 on Build GRCh38
1081576 c.900A>C p.K300N Chr.6: 73762785-73762785 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751932439 Chr.6: 73762837-73762837 on Build GRCh38
rs999329170 Chr.6: 73762814-73762814 on Build GRCh38
rs993720198 Chr.6: 73762677-73762677 on Build GRCh38
rs778560293 Chr.6: 73762799-73762799 on Build GRCh38
rs1037834278 Chr.6: 73762667-73762667 on Build GRCh38
rs759223728 Chr.6: 73762653-73762653 on Build GRCh38
rs374417084 Chr.6: 73762742-73762742 on Build GRCh38
rs775042813 Chr.6: 73762816-73762816 on Build GRCh38
rs764634866 Chr.6: 73762714-73762714 on Build GRCh38
rs770996662 Chr.6: 73762796-73762796 on Build GRCh38
rs768849691 Chr.6: 73762832-73762832 on Build GRCh38
rs745425108 Chr.6: 73762802-73762802 on Build GRCh38
rs1018104990 Chr.6: 73762780-73762780 on Build GRCh38
rs760402885 Chr.6: 73762830-73762830 on Build GRCh38
rs914378428 Chr.6: 73762820-73762820 on Build GRCh38
rs964788422 Chr.6: 73762789-73762789 on Build GRCh38
rs760997858 Chr.6: 73762709-73762709 on Build GRCh38
rs754306739 Chr.6: 73762717-73762717 on Build GRCh38
rs975227334 Chr.6: 73762803-73762803 on Build GRCh38
rs567645940 Chr.6: 73762781-73762781 on Build GRCh38
rs899421270 Chr.6: 73762676-73762676 on Build GRCh38
rs762244483 Chr.6: 73762835-73762835 on Build GRCh38
rs750510625 Chr.6: 73762756-73762756 on Build GRCh38
rs928381842 Chr.6: 73762665-73762665 on Build GRCh38
rs529156331 Chr.6: 73762686-73762686 on Build GRCh38
rs775911828 Chr.6: 73762702-73762702 on Build GRCh38
rs749179661 Chr.6: 73762795-73762795 on Build GRCh38
rs2351312 Chr.6: 73762768-73762768 on Build GRCh38
rs765802724 Chr.6: 73762838-73762838 on Build GRCh38
rs765239774 Chr.6: 73762747-73762747 on Build GRCh38
rs1043278786 Chr.6: 73762778-73762778 on Build GRCh38
rs200666210 Chr.6: 73762808-73762808 on Build GRCh38
rs368503067 Chr.6: 73762773-73762773 on Build GRCh38
rs547286744 Chr.6: 73762777-73762777 on Build GRCh38
rs780200187 Chr.6: 73762776-73762776 on Build GRCh38
rs968625641 Chr.6: 73762821-73762821 on Build GRCh38

View Full Product Details