Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6652202-6652475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ING4
Entrez Gene ID
51147
Gene Name
inhibitor of growth family member 4
Gene Aliases
my036, p29ING4
UniGene
Hs.524210
Gene Chromosome Location
Chr.12: 6650538-6663143 on Build GRCh38
Transcripts
NM_001127582.1 NM_001127583.1 NM_001127584.1 NM_001127585.1 NM_001127586.1 NM_016162.3 XM_005253698.3 XM_011520964.2 XM_011520965.2 XM_017019392.1 XM_017019393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6137880 c.629G>T p.G210V Chr.12: 6652287-6652287 on Build GRCh38
4502495 c.611C>T p.S204F Chr.12: 6652305-6652305 on Build GRCh38
4575681 c.519C>T p.P173P Chr.12: 6652397-6652397 on Build GRCh38
344202 c.498-2A>T p.? Chr.12: 6652420-6652420 on Build GRCh38
4554976 c.634G>C p.D212H Chr.12: 6652282-6652282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760396496 Chr.12: 6652331-6652331 on Build GRCh38
rs775709391 Chr.12: 6652415-6652415 on Build GRCh38
rs536793678 Chr.12: 6652453-6652453 on Build GRCh38
rs759537062 Chr.12: 6652233-6652233 on Build GRCh38
rs201257362 Chr.12: 6652279-6652279 on Build GRCh38
rs771207486 Chr.12: 6652400-6652400 on Build GRCh38
rs763876954 Chr.12: 6652262-6652262 on Build GRCh38
rs146347929 Chr.12: 6652397-6652397 on Build GRCh38
rs757869141 Chr.12: 6652305-6652305 on Build GRCh38
rs768597105 Chr.12: 6652442-6652442 on Build GRCh38
rs776346199 Chr.12: 6652444-6652444 on Build GRCh38
rs536407343 Chr.12: 6652451-6652451 on Build GRCh38
rs77283008 Chr.12: 6652271-6652271 on Build GRCh38
rs749911348 Chr.12: 6652284-6652284 on Build GRCh38
rs756751323 Chr.12: 6652338-6652338 on Build GRCh38
rs762652667 Chr.12: 6652429-6652429 on Build GRCh38
rs778267798 Chr.12: 6652373-6652373 on Build GRCh38
rs565994242 Chr.12: 6652234-6652234 on Build GRCh38
rs371654809 Chr.12: 6652403-6652403 on Build GRCh38
rs772019749 Chr.12: 6652414-6652414 on Build GRCh38
rs867811925 Chr.12: 6652417-6652417 on Build GRCh38
rs774512004 Chr.12: 6652230-6652230 on Build GRCh38
rs369007352 Chr.12: 6652310-6652310 on Build GRCh38
rs745907388 Chr.12: 6652413-6652413 on Build GRCh38
rs1051146228 Chr.12: 6652389-6652389 on Build GRCh38
rs113711890 Chr.12: 6652419-6652419 on Build GRCh38
rs377231817 Chr.12: 6652432-6652432 on Build GRCh38
rs779664155 Chr.12: 6652316-6652316 on Build GRCh38
rs369199001 Chr.12: 6652241-6652241 on Build GRCh38
rs760710150 Chr.12: 6652243-6652243 on Build GRCh38
rs761628715 Chr.12: 6652450-6652450 on Build GRCh38
rs375300631 Chr.12: 6652246-6652246 on Build GRCh38
rs867902357 Chr.12: 6652311-6652311 on Build GRCh38
rs756848334 Chr.12: 6652276-6652276 on Build GRCh38
rs777485323 Chr.12: 6652244-6652244 on Build GRCh38
rs769576351 Chr.12: 6652278-6652278 on Build GRCh38
rs748571571 Chr.12: 6652337-6652337 on Build GRCh38

View Full Product Details