Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136339674-136339937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPSM1
Entrez Gene ID
26086
Gene Name
G-protein signaling modulator 1
Gene Aliases
AGS3
UniGene
Hs.239370
Gene Chromosome Location
Chr.9: 136327476-136359605 on Build GRCh38
Transcripts
NM_001145638.2 NM_015597.5 XM_011518498.2 XM_017014598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500239 c.940A>T p.N314Y Chr.9: 136339741-136339741 on Build GRCh38
4936737 c.1071G>A p.Q357Q Chr.9: 136339803-136339803 on Build GRCh38
6418079 c.1051T>C p.F351L Chr.9: 136339783-136339783 on Build GRCh38
5608973 c.1008C>T p.S336S Chr.9: 136339809-136339809 on Build GRCh38
3367639 c.910G>A p.G304S Chr.9: 136339711-136339711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368271090 Chr.9: 136339898-136339898 on Build GRCh38
rs551509416 Chr.9: 136339810-136339810 on Build GRCh38
rs34940813 Chr.9: 136339903-136339904 on Build GRCh38
rs782148747 Chr.9: 136339861-136339861 on Build GRCh38
rs782810238 Chr.9: 136339868-136339868 on Build GRCh38
rs782078428 Chr.9: 136339770-136339770 on Build GRCh38
rs549086721 Chr.9: 136339914-136339914 on Build GRCh38
rs948375501 Chr.9: 136339884-136339884 on Build GRCh38
rs969241759 Chr.9: 136339805-136339805 on Build GRCh38
rs199771282 Chr.9: 136339804-136339804 on Build GRCh38
rs530589273 Chr.9: 136339874-136339874 on Build GRCh38
rs924471732 Chr.9: 136339830-136339830 on Build GRCh38
rs375591793 Chr.9: 136339777-136339777 on Build GRCh38
rs977279754 Chr.9: 136339812-136339812 on Build GRCh38
rs781937612 Chr.9: 136339720-136339720 on Build GRCh38
rs141313816 Chr.9: 136339749-136339749 on Build GRCh38
rs782781438 Chr.9: 136339765-136339765 on Build GRCh38
rs372088202 Chr.9: 136339713-136339713 on Build GRCh38
rs373488186 Chr.9: 136339769-136339769 on Build GRCh38
rs782553393 Chr.9: 136339844-136339844 on Build GRCh38
rs997255811 Chr.9: 136339785-136339785 on Build GRCh38
rs369402356 Chr.9: 136339762-136339762 on Build GRCh38
rs146990496 Chr.9: 136339760-136339760 on Build GRCh38
rs1037782858 Chr.9: 136339894-136339894 on Build GRCh38
rs115335934 Chr.9: 136339869-136339869 on Build GRCh38

View Full Product Details