Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196215750-196216023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5742898 c.4528G>T p.G1510* Chr.2: 196215944-196215944 on Build GRCh38
6678934 c.4554C>T p.N1518N Chr.2: 196215918-196215918 on Build GRCh38
5928294 c.4605C>T p.P1535P Chr.2: 196215867-196215867 on Build GRCh38
418937 c.4587G>T p.G1529G Chr.2: 196215885-196215885 on Build GRCh38
5465346 c.4532T>G p.F1511C Chr.2: 196215940-196215940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777318950 Chr.2: 196215840-196215840 on Build GRCh38
rs963429357 Chr.2: 196215907-196215907 on Build GRCh38
rs771541458 Chr.2: 196215839-196215839 on Build GRCh38
rs750792513 Chr.2: 196215946-196215946 on Build GRCh38
rs778094004 Chr.2: 196215771-196215771 on Build GRCh38
rs376055248 Chr.2: 196215885-196215885 on Build GRCh38
rs908966016 Chr.2: 196215847-196215847 on Build GRCh38
rs749797656 Chr.2: 196215837-196215837 on Build GRCh38
rs757448581 Chr.2: 196215817-196215817 on Build GRCh38
rs749523740 Chr.2: 196215791-196215791 on Build GRCh38
rs771850080 Chr.2: 196215852-196215852 on Build GRCh38
rs780453901 Chr.2: 196215970-196215970 on Build GRCh38
rs770669991 Chr.2: 196215882-196215882 on Build GRCh38
rs778845144 Chr.2: 196215836-196215836 on Build GRCh38
rs866202822 Chr.2: 196215926-196215926 on Build GRCh38
rs9288259 Chr.2: 196215830-196215830 on Build GRCh38
rs756538096 Chr.2: 196215968-196215968 on Build GRCh38
rs776586750 Chr.2: 196215884-196215884 on Build GRCh38
rs940001217 Chr.2: 196215889-196215889 on Build GRCh38
rs149233137 Chr.2: 196215912-196215912 on Build GRCh38
rs754354742 Chr.2: 196215982-196215982 on Build GRCh38
rs746684990 Chr.2: 196215876-196215876 on Build GRCh38
rs750745354 Chr.2: 196215828-196215828 on Build GRCh38
rs769855164 Chr.2: 196215888-196215888 on Build GRCh38
rs781574219 Chr.2: 196215825-196215825 on Build GRCh38
rs767670181 Chr.2: 196215930-196215930 on Build GRCh38
rs545923415 Chr.2: 196215894-196215894 on Build GRCh38
rs559297514 Chr.2: 196215899-196215899 on Build GRCh38
rs17310196 Chr.2: 196215918-196215918 on Build GRCh38
rs762051916 Chr.2: 196215928-196215928 on Build GRCh38

View Full Product Details