Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79781457-79781728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX2
Entrez Gene ID
84733
Gene Name
chromobox 2
Gene Aliases
CDCA6, M33, SRXY5
UniGene
Hs.368410
Gene Chromosome Location
Chr.17: 79776254-79787650 on Build GRCh38
Transcripts
NM_005189.2 NM_032647.3 XM_011525382.1 XM_011525383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423087 c.184G>T p.E62* Chr.17: 79781697-79781697 on Build GRCh38
7423088 c.184G>T p.E62* Chr.17: 79781697-79781697 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781984713 Chr.17: 79781653-79781653 on Build GRCh38
rs782260786 Chr.17: 79781654-79781654 on Build GRCh38
rs185563278 Chr.17: 79781639-79781639 on Build GRCh38
rs782455267 Chr.17: 79781697-79781697 on Build GRCh38
rs567947932 Chr.17: 79781642-79781642 on Build GRCh38
rs782720820 Chr.17: 79781669-79781669 on Build GRCh38
rs782769415 Chr.17: 79781674-79781674 on Build GRCh38
rs996212645 Chr.17: 79781554-79781554 on Build GRCh38
rs781788632 Chr.17: 79781693-79781693 on Build GRCh38
rs782798940 Chr.17: 79781690-79781690 on Build GRCh38
rs782405022 Chr.17: 79781655-79781655 on Build GRCh38
rs978297495 Chr.17: 79781590-79781590 on Build GRCh38
rs1018675240 Chr.17: 79781518-79781518 on Build GRCh38
rs559963323 Chr.17: 79781521-79781521 on Build GRCh38
rs782600574 Chr.17: 79781699-79781699 on Build GRCh38
rs782037228 Chr.17: 79781658-79781658 on Build GRCh38
rs781863293 Chr.17: 79781685-79781685 on Build GRCh38
rs781957109 Chr.17: 79781671-79781671 on Build GRCh38
rs782248219 Chr.17: 79781708-79781708 on Build GRCh38
rs1038808593 Chr.17: 79781665-79781665 on Build GRCh38
rs373920215 Chr.17: 79781688-79781688 on Build GRCh38
rs782108761 Chr.17: 79781673-79781673 on Build GRCh38
rs115374900 Chr.17: 79781575-79781575 on Build GRCh38
rs775175047 Chr.17: 79781545-79781545 on Build GRCh38
rs202115702 Chr.17: 79781668-79781668 on Build GRCh38
rs552106640 Chr.17: 79781576-79781576 on Build GRCh38
rs919933175 Chr.17: 79781666-79781666 on Build GRCh38
rs570103121 Chr.17: 79781611-79781611 on Build GRCh38
rs547810772 Chr.17: 79781520-79781520 on Build GRCh38
rs960833976 Chr.17: 79781610-79781610 on Build GRCh38
rs181291524 Chr.17: 79781629-79781629 on Build GRCh38
rs930570771 Chr.17: 79781704-79781704 on Build GRCh38
rs1008679997 Chr.17: 79781511-79781511 on Build GRCh38
rs924143250 Chr.17: 79781594-79781594 on Build GRCh38
rs115837021 Chr.17: 79781643-79781643 on Build GRCh38
rs375104924 Chr.17: 79781663-79781663 on Build GRCh38

View Full Product Details