Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44558603-44558852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD2
Entrez Gene ID
2535
Gene Name
frizzled class receptor 2
Gene Aliases
Fz2, fz-2, fzE2, hFz2
UniGene
Hs.142912
Gene Chromosome Location
Chr.17: 44557444-44561262 on Build GRCh38
Transcripts
NM_001466.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6041260 c.1046C>T p.S349L Chr.17: 44558734-44558734 on Build GRCh38
1128686 c.1005C>G p.Y335* Chr.17: 44558693-44558693 on Build GRCh38
1128684 c.1117C>T p.H373Y Chr.17: 44558805-44558805 on Build GRCh38
5852666 c.934G>A p.E312K Chr.17: 44558622-44558622 on Build GRCh38
1128685 c.1047G>A p.S349S Chr.17: 44558735-44558735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925950278 Chr.17: 44558658-44558658 on Build GRCh38
rs755024659 Chr.17: 44558783-44558783 on Build GRCh38
rs574279557 Chr.17: 44558633-44558633 on Build GRCh38
rs764576821 Chr.17: 44558656-44558656 on Build GRCh38
rs530531830 Chr.17: 44558829-44558829 on Build GRCh38
rs866732079 Chr.17: 44558710-44558710 on Build GRCh38
rs777369808 Chr.17: 44558806-44558806 on Build GRCh38
rs144700537 Chr.17: 44558825-44558825 on Build GRCh38
rs756235284 Chr.17: 44558756-44558756 on Build GRCh38
rs750684480 Chr.17: 44558729-44558729 on Build GRCh38
rs778973007 Chr.17: 44558785-44558785 on Build GRCh38
rs767549516 Chr.17: 44558715-44558715 on Build GRCh38
rs901265781 Chr.17: 44558735-44558735 on Build GRCh38
rs267604907 Chr.17: 44558714-44558714 on Build GRCh38
rs367593562 Chr.17: 44558774-44558774 on Build GRCh38
rs775656853 Chr.17: 44558627-44558627 on Build GRCh38
rs563435543 Chr.17: 44558801-44558801 on Build GRCh38
rs1049538268 Chr.17: 44558828-44558828 on Build GRCh38
rs770215499 Chr.17: 44558622-44558622 on Build GRCh38
rs747701116 Chr.17: 44558791-44558791 on Build GRCh38
rs374157449 Chr.17: 44558771-44558771 on Build GRCh38
rs1043290843 Chr.17: 44558640-44558640 on Build GRCh38
rs879493296 Chr.17: 44558822-44558822 on Build GRCh38
rs373390114 Chr.17: 44558666-44558666 on Build GRCh38
rs1053647443 Chr.17: 44558687-44558687 on Build GRCh38
rs746685961 Chr.17: 44558823-44558823 on Build GRCh38
rs866062060 Chr.17: 44558792-44558792 on Build GRCh38

View Full Product Details