Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44057759-44058026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBNL
Entrez Gene ID
28988
Gene Name
drebrin like
Gene Aliases
ABP1, HIP-55, HIP55, SH3P7
UniGene
Hs.436500
Gene Chromosome Location
Chr.7: 44044640-44061716 on Build GRCh38
Transcripts
NM_001014436.2 NM_001122956.1 NM_001284313.1 NM_001284315.1 NM_014063.6 XM_011515325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272579 c.540G>T p.W180C Chr.7: 44057847-44057847 on Build GRCh38
4676295 c.546A>C p.K182N Chr.7: 44057853-44057853 on Build GRCh38
6272580 c.540G>T p.W180C Chr.7: 44057847-44057847 on Build GRCh38
6504616 c.503T>C p.V168A Chr.7: 44057810-44057810 on Build GRCh38
3929051 c.545A>G p.K182R Chr.7: 44057852-44057852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763040275 Chr.7: 44057862-44057862 on Build GRCh38
rs778262161 Chr.7: 44057909-44057909 on Build GRCh38
rs751475220 Chr.7: 44057873-44057873 on Build GRCh38
rs544660124 Chr.7: 44057983-44057984 on Build GRCh38
rs888783600 Chr.7: 44057942-44057943 on Build GRCh38
rs774834305 Chr.7: 44057792-44057792 on Build GRCh38
rs961991441 Chr.7: 44057982-44057982 on Build GRCh38
rs917206743 Chr.7: 44057874-44057874 on Build GRCh38
rs752801449 Chr.7: 44057851-44057851 on Build GRCh38
rs761036087 Chr.7: 44057814-44057814 on Build GRCh38
rs771138867 Chr.7: 44057820-44057820 on Build GRCh38
rs944427711 Chr.7: 44057963-44057963 on Build GRCh38
rs370531550 Chr.7: 44057889-44057889 on Build GRCh38
rs202107637 Chr.7: 44057886-44057886 on Build GRCh38
rs773636683 Chr.7: 44057809-44057809 on Build GRCh38
rs750132157 Chr.7: 44057885-44057885 on Build GRCh38
rs1018627117 Chr.7: 44057949-44057949 on Build GRCh38
rs759635060 Chr.7: 44057830-44057830 on Build GRCh38
rs138446917 Chr.7: 44057796-44057796 on Build GRCh38
rs112056427 Chr.7: 44057860-44057860 on Build GRCh38
rs776930553 Chr.7: 44057828-44057828 on Build GRCh38
rs780811051 Chr.7: 44057881-44057881 on Build GRCh38
rs757098575 Chr.7: 44057876-44057876 on Build GRCh38
rs141349639 Chr.7: 44057841-44057841 on Build GRCh38
rs971842235 Chr.7: 44057990-44057990 on Build GRCh38
rs768091895 Chr.7: 44057892-44057892 on Build GRCh38
rs748705979 Chr.7: 44057795-44057795 on Build GRCh38
rs564758049 Chr.7: 44057871-44057871 on Build GRCh38
rs547192374 Chr.7: 44057991-44057991 on Build GRCh38
rs769378674 Chr.7: 44057786-44057786 on Build GRCh38
rs368023575 Chr.7: 44057888-44057888 on Build GRCh38
rs1050102119 Chr.7: 44057923-44057923 on Build GRCh38
rs753874693 Chr.7: 44057981-44057981 on Build GRCh38

View Full Product Details