Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75335993-75336242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD4
Entrez Gene ID
54939
Gene Name
COMM domain containing 4
Gene Aliases
-
UniGene
Hs.351327
Gene Chromosome Location
Chr.15: 75335996-75341927 on Build GRCh38
Transcripts
NM_001284377.1 NM_001284378.1 NM_001284379.1 NM_001321844.1 NM_001321845.1 NM_001321846.1 NM_001321847.1 NM_001321848.1 NM_017828.4 NR_104312.1 NR_104313.1 XM_005254509.3 XM_005254510.3 XM_011521739.1 XM_011521740.1 XM_011521741.1 XM_017022385.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608686 c.3+1G>A p.? Chr.15: 75336093-75336093 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369533129 Chr.15: 75336119-75336119 on Build GRCh38
rs779218903 Chr.15: 75336183-75336183 on Build GRCh38
rs563232982 Chr.15: 75336201-75336201 on Build GRCh38
rs928523199 Chr.15: 75336067-75336067 on Build GRCh38
rs983758724 Chr.15: 75336185-75336185 on Build GRCh38
rs768714546 Chr.15: 75336113-75336113 on Build GRCh38
rs893109278 Chr.15: 75336098-75336098 on Build GRCh38
rs567672022 Chr.15: 75336053-75336053 on Build GRCh38
rs772757356 Chr.15: 75336149-75336149 on Build GRCh38
rs765045060 Chr.15: 75336072-75336072 on Build GRCh38
rs186098733 Chr.15: 75336071-75336071 on Build GRCh38
rs865974685 Chr.15: 75336152-75336152 on Build GRCh38
rs759900365 Chr.15: 75336099-75336099 on Build GRCh38
rs751270022 Chr.15: 75336082-75336082 on Build GRCh38
rs770893626 Chr.15: 75336151-75336151 on Build GRCh38
rs1015954010 Chr.15: 75336200-75336201 on Build GRCh38
rs764813364 Chr.15: 75336211-75336211 on Build GRCh38
rs539928757 Chr.15: 75336102-75336102 on Build GRCh38
rs372819742 Chr.15: 75336069-75336069 on Build GRCh38
rs370496699 Chr.15: 75336065-75336065 on Build GRCh38
rs749680697 Chr.15: 75336057-75336057 on Build GRCh38
rs775559665 Chr.15: 75336091-75336091 on Build GRCh38
rs757224491 Chr.15: 75336090-75336090 on Build GRCh38
rs867868649 Chr.15: 75336068-75336068 on Build GRCh38
rs557643509 Chr.15: 75336080-75336080 on Build GRCh38
rs1053154607 Chr.15: 75336093-75336093 on Build GRCh38
rs114064595 Chr.15: 75336073-75336073 on Build GRCh38
rs764194546 Chr.15: 75336063-75336063 on Build GRCh38
rs781278530 Chr.15: 75336043-75336043 on Build GRCh38
rs767869554 Chr.15: 75336126-75336126 on Build GRCh38
rs773847543 Chr.15: 75336178-75336178 on Build GRCh38
rs752340100 Chr.15: 75336115-75336115 on Build GRCh38
rs770048903 Chr.15: 75336051-75336051 on Build GRCh38
rs932776208 Chr.15: 75336077-75336077 on Build GRCh38
rs182080358 Chr.15: 75336066-75336066 on Build GRCh38
rs543831475 Chr.15: 75336116-75336116 on Build GRCh38
rs377456117 Chr.15: 75336184-75336184 on Build GRCh38
rs891417854 Chr.15: 75336086-75336086 on Build GRCh38
rs371912035 Chr.15: 75336108-75336108 on Build GRCh38
rs572646649 Chr.15: 75336100-75336100 on Build GRCh38
rs745916276 Chr.15: 75336044-75336044 on Build GRCh38
rs781104421 Chr.15: 75336051-75336051 on Build GRCh38
rs749428321 Chr.15: 75336087-75336087 on Build GRCh38
rs555838912 Chr.15: 75336030-75336030 on Build GRCh38
rs770803285 Chr.15: 75336041-75336041 on Build GRCh38
rs769069935 Chr.15: 75336060-75336060 on Build GRCh38
rs749539025 Chr.15: 75336112-75336112 on Build GRCh38
rs367746601 Chr.15: 75336107-75336107 on Build GRCh38

View Full Product Details