Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149750763-149750990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD2
Entrez Gene ID
51122
Gene Name
COMM domain containing 2
Gene Aliases
HSPC042
UniGene
Hs.432729
Gene Chromosome Location
Chr.3: 149737160-149752507 on Build GRCh38
Transcripts
NM_016094.3 NR_135149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3846493 c.280G>T p.E94* Chr.3: 149750800-149750800 on Build GRCh38
7316190 c.310C>G p.L104V Chr.3: 149750770-149750770 on Build GRCh38
3915115 c.244T>C p.F82L Chr.3: 149750836-149750836 on Build GRCh38
5431961 c.289A>C p.K97Q Chr.3: 149750791-149750791 on Build GRCh38
5803872 c.282A>G p.E94E Chr.3: 149750798-149750798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771993406 Chr.3: 149750827-149750827 on Build GRCh38
rs749538024 Chr.3: 149750797-149750797 on Build GRCh38
rs772114958 Chr.3: 149750825-149750826 on Build GRCh38
rs993350782 Chr.3: 149750908-149750908 on Build GRCh38
rs771247836 Chr.3: 149750805-149750805 on Build GRCh38
rs75480034 Chr.3: 149750893-149750893 on Build GRCh38
rs887723448 Chr.3: 149750930-149750930 on Build GRCh38
rs776007210 Chr.3: 149750856-149750856 on Build GRCh38
rs761101407 Chr.3: 149750876-149750876 on Build GRCh38
rs1036671868 Chr.3: 149750928-149750928 on Build GRCh38
rs775518020 Chr.3: 149750871-149750872 on Build GRCh38
rs754259318 Chr.3: 149750887-149750887 on Build GRCh38
rs112243276 Chr.3: 149750837-149750837 on Build GRCh38
rs200102094 Chr.3: 149750898-149750898 on Build GRCh38
rs200273536 Chr.3: 149750812-149750812 on Build GRCh38
rs376996550 Chr.3: 149750862-149750862 on Build GRCh38
rs980019090 Chr.3: 149750847-149750847 on Build GRCh38
rs760576903 Chr.3: 149750901-149750901 on Build GRCh38
rs1006219206 Chr.3: 149750962-149750962 on Build GRCh38
rs764455976 Chr.3: 149750886-149750886 on Build GRCh38
rs762081259 Chr.3: 149750891-149750891 on Build GRCh38
rs113167845 Chr.3: 149750894-149750894 on Build GRCh38

View Full Product Details