Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36357177-36357446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895
Entrez Gene ID
23366
Gene Name
KIAA0895
Gene Aliases
-
UniGene
Hs.6224
Gene Chromosome Location
Chr.7: 36324150-36390125 on Build GRCh38
Transcripts
NM_001100425.1 NM_001199706.1 NM_001199707.1 NM_001199708.1 NM_001300956.1 NM_015314.2 XM_005249689.3 XM_005249690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4548929 c.460G>A p.A154T Chr.7: 36357309-36357309 on Build GRCh38
6109909 c.319G>T p.V107L Chr.7: 36357297-36357297 on Build GRCh38
3414012 c.231C>T p.S77S Chr.7: 36357385-36357385 on Build GRCh38
7261263 c.409C>T p.R137C Chr.7: 36357321-36357321 on Build GRCh38
485293 c.492G>T p.W164C Chr.7: 36357277-36357277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775965678 Chr.7: 36357368-36357368 on Build GRCh38
rs201511349 Chr.7: 36357336-36357336 on Build GRCh38
rs751759163 Chr.7: 36357261-36357261 on Build GRCh38
rs369116622 Chr.7: 36357215-36357215 on Build GRCh38
rs750555847 Chr.7: 36357403-36357403 on Build GRCh38
rs200135558 Chr.7: 36357404-36357404 on Build GRCh38
rs777338392 Chr.7: 36357346-36357348 on Build GRCh38
rs757310600 Chr.7: 36357272-36357272 on Build GRCh38
rs78387918 Chr.7: 36357252-36357252 on Build GRCh38
rs369837787 Chr.7: 36357416-36357416 on Build GRCh38
rs765631482 Chr.7: 36357202-36357202 on Build GRCh38
rs763335153 Chr.7: 36357381-36357381 on Build GRCh38
rs113184966 Chr.7: 36357320-36357320 on Build GRCh38
rs781260158 Chr.7: 36357282-36357282 on Build GRCh38
rs749160285 Chr.7: 36357317-36357317 on Build GRCh38
rs756106791 Chr.7: 36357308-36357308 on Build GRCh38
rs753754055 Chr.7: 36357413-36357413 on Build GRCh38
rs201268339 Chr.7: 36357220-36357220 on Build GRCh38
rs1009859024 Chr.7: 36357369-36357369 on Build GRCh38
rs771651088 Chr.7: 36357335-36357335 on Build GRCh38
rs779816032 Chr.7: 36357316-36357316 on Build GRCh38
rs756027269 Chr.7: 36357409-36357409 on Build GRCh38
rs747780074 Chr.7: 36357333-36357333 on Build GRCh38
rs764386192 Chr.7: 36357384-36357384 on Build GRCh38
rs778590332 Chr.7: 36357326-36357326 on Build GRCh38
rs760116958 Chr.7: 36357343-36357343 on Build GRCh38
rs780010491 Chr.7: 36357412-36357412 on Build GRCh38
rs369438919 Chr.7: 36357297-36357297 on Build GRCh38
rs199901322 Chr.7: 36357260-36357260 on Build GRCh38
rs377702299 Chr.7: 36357393-36357393 on Build GRCh38
rs1021216863 Chr.7: 36357377-36357377 on Build GRCh38
rs770471733 Chr.7: 36357352-36357352 on Build GRCh38
rs767576321 Chr.7: 36357395-36357395 on Build GRCh38
rs751744428 Chr.7: 36357388-36357388 on Build GRCh38

View Full Product Details