Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45543468-45543727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN1
Entrez Gene ID
2192
Gene Name
fibulin 1
Gene Aliases
FBLN, FIBL1
UniGene
Hs.24601
Gene Chromosome Location
Chr.22: 45502839-45601134 on Build GRCh38
Transcripts
NM_001996.3 NM_006485.3 NM_006486.2 NM_006487.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5393834 c.1269C>G p.L423L Chr.22: 45543474-45543474 on Build GRCh38
1682327 c.1291C>T p.R431W Chr.22: 45543496-45543496 on Build GRCh38
1682328 c.1291C>T p.R431W Chr.22: 45543496-45543496 on Build GRCh38
5095893 c.1313C>A p.S438* Chr.22: 45543518-45543518 on Build GRCh38
4300499 c.1291C>T p.R431W Chr.22: 45543496-45543496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186612266 Chr.22: 45543657-45543657 on Build GRCh38
rs1023830530 Chr.22: 45543701-45543701 on Build GRCh38
rs926074361 Chr.22: 45543600-45543600 on Build GRCh38
rs892801819 Chr.22: 45543678-45543678 on Build GRCh38
rs769192815 Chr.22: 45543518-45543518 on Build GRCh38
rs573887808 Chr.22: 45543536-45543536 on Build GRCh38
rs16994215 Chr.22: 45543616-45543616 on Build GRCh38
rs746761995 Chr.22: 45543564-45543564 on Build GRCh38
rs574765946 Chr.22: 45543537-45543537 on Build GRCh38
rs746921355 Chr.22: 45543501-45543501 on Build GRCh38
rs962459918 Chr.22: 45543563-45543563 on Build GRCh38
rs934244861 Chr.22: 45543668-45543668 on Build GRCh38
rs757513972 Chr.22: 45543555-45543555 on Build GRCh38
rs920599820 Chr.22: 45543569-45543569 on Build GRCh38
rs771837629 Chr.22: 45543497-45543497 on Build GRCh38
rs112157355 Chr.22: 45543694-45543694 on Build GRCh38
rs545983411 Chr.22: 45543618-45543618 on Build GRCh38
rs773192652 Chr.22: 45543535-45543535 on Build GRCh38
rs577073351 Chr.22: 45543617-45543617 on Build GRCh38
rs779141731 Chr.22: 45543559-45543559 on Build GRCh38
rs902908546 Chr.22: 45543653-45543653 on Build GRCh38
rs776459410 Chr.22: 45543506-45543506 on Build GRCh38
rs369516282 Chr.22: 45543509-45543509 on Build GRCh38
rs113497852 Chr.22: 45543527-45543527 on Build GRCh38
rs771043632 Chr.22: 45543644-45543644 on Build GRCh38
rs905468263 Chr.22: 45543704-45543704 on Build GRCh38
rs768009104 Chr.22: 45543543-45543543 on Build GRCh38
rs562408229 Chr.22: 45543621-45543621 on Build GRCh38
rs934631524 Chr.22: 45543613-45543613 on Build GRCh38
rs777982991 Chr.22: 45543495-45543495 on Build GRCh38
rs753145561 Chr.22: 45543546-45543546 on Build GRCh38
rs969577444 Chr.22: 45543524-45543524 on Build GRCh38
rs200614579 Chr.22: 45543487-45543487 on Build GRCh38
rs963030598 Chr.22: 45543498-45543498 on Build GRCh38
rs372500771 Chr.22: 45543552-45543552 on Build GRCh38
rs756210058 Chr.22: 45543547-45543547 on Build GRCh38
rs371708304 Chr.22: 45543591-45543591 on Build GRCh38
rs915733234 Chr.22: 45543568-45543568 on Build GRCh38
rs766818305 Chr.22: 45543540-45543540 on Build GRCh38
rs765649784 Chr.22: 45543551-45543551 on Build GRCh38
rs768544289 Chr.22: 45543505-45543505 on Build GRCh38
rs778458730 Chr.22: 45543542-45543542 on Build GRCh38
rs772915964 Chr.22: 45543519-45543519 on Build GRCh38
rs1054010740 Chr.22: 45543669-45543669 on Build GRCh38
rs747782991 Chr.22: 45543583-45543583 on Build GRCh38
rs749537377 Chr.22: 45543496-45543496 on Build GRCh38
rs771770589 Chr.22: 45543599-45543599 on Build GRCh38
rs1050452915 Chr.22: 45543673-45543673 on Build GRCh38
rs764640380 Chr.22: 45543606-45543606 on Build GRCh38
rs910505052 Chr.22: 45543615-45543615 on Build GRCh38
rs561392327 Chr.22: 45543703-45543703 on Build GRCh38
rs754783081 Chr.22: 45543574-45543574 on Build GRCh38
rs972027739 Chr.22: 45543567-45543567 on Build GRCh38
rs982718683 Chr.22: 45543533-45543533 on Build GRCh38

View Full Product Details