Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6217257-6217504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5880361 c.2140_2141insG p.L714fs*4 Chr.11: 6217487-6217488 on Build GRCh38
6133716 c.2247G>C p.Q749H Chr.11: 6217381-6217381 on Build GRCh38
1355325 c.2195delC p.P732fs*46 Chr.11: 6217433-6217433 on Build GRCh38
6658704 c.2215C>T p.P739S Chr.11: 6217413-6217413 on Build GRCh38
5562880 c.2183C>T p.P728L Chr.11: 6217445-6217445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148057736 Chr.11: 6217406-6217406 on Build GRCh38
rs746934506 Chr.11: 6217358-6217358 on Build GRCh38
rs776428818 Chr.11: 6217397-6217397 on Build GRCh38
rs967763606 Chr.11: 6217386-6217386 on Build GRCh38
rs770556663 Chr.11: 6217283-6217283 on Build GRCh38
rs568595635 Chr.11: 6217315-6217315 on Build GRCh38
rs986982424 Chr.11: 6217455-6217455 on Build GRCh38
rs758886166 Chr.11: 6217320-6217320 on Build GRCh38
rs202172236 Chr.11: 6217461-6217461 on Build GRCh38
rs115637949 Chr.11: 6217305-6217305 on Build GRCh38
rs753131011 Chr.11: 6217437-6217437 on Build GRCh38
rs781243507 Chr.11: 6217477-6217477 on Build GRCh38
rs775217953 Chr.11: 6217347-6217347 on Build GRCh38
rs771892659 Chr.11: 6217345-6217345 on Build GRCh38
rs768573102 Chr.11: 6217381-6217381 on Build GRCh38
rs751673440 Chr.11: 6217301-6217301 on Build GRCh38
rs757522136 Chr.11: 6217441-6217442 on Build GRCh38
rs972396087 Chr.11: 6217436-6217436 on Build GRCh38
rs748760819 Chr.11: 6217330-6217330 on Build GRCh38
rs765388868 Chr.11: 6217401-6217401 on Build GRCh38
rs778593557 Chr.11: 6217341-6217341 on Build GRCh38
rs72910144 Chr.11: 6217433-6217433 on Build GRCh38
rs140870715 Chr.11: 6217450-6217450 on Build GRCh38
rs370706455 Chr.11: 6217327-6217327 on Build GRCh38
rs534028611 Chr.11: 6217374-6217374 on Build GRCh38
rs775453306 Chr.11: 6217298-6217298 on Build GRCh38
rs755460531 Chr.11: 6217329-6217329 on Build GRCh38
rs956377446 Chr.11: 6217335-6217335 on Build GRCh38
rs887864890 Chr.11: 6217296-6217296 on Build GRCh38
rs774136778 Chr.11: 6217287-6217287 on Build GRCh38
rs141812318 Chr.11: 6217407-6217407 on Build GRCh38
rs374871287 Chr.11: 6217442-6217442 on Build GRCh38
rs781090912 Chr.11: 6217449-6217454 on Build GRCh38
rs773612493 Chr.11: 6217403-6217403 on Build GRCh38
rs1049096279 Chr.11: 6217478-6217478 on Build GRCh38
rs755441126 Chr.11: 6217428-6217428 on Build GRCh38
rs779830271 Chr.11: 6217446-6217446 on Build GRCh38
rs764105374 Chr.11: 6217301-6217301 on Build GRCh38
rs778360593 Chr.11: 6217438-6217438 on Build GRCh38
rs529000908 Chr.11: 6217290-6217290 on Build GRCh38
rs751891419 Chr.11: 6217412-6217412 on Build GRCh38
rs762015454 Chr.11: 6217399-6217399 on Build GRCh38
rs745497207 Chr.11: 6217344-6217344 on Build GRCh38
rs762421923 Chr.11: 6217288-6217288 on Build GRCh38
rs756717084 Chr.11: 6217340-6217340 on Build GRCh38
rs765565992 Chr.11: 6217319-6217319 on Build GRCh38
rs1021969443 Chr.11: 6217363-6217363 on Build GRCh38
rs962349630 Chr.11: 6217434-6217434 on Build GRCh38
rs141597566 Chr.11: 6217484-6217484 on Build GRCh38
rs748912813 Chr.11: 6217279-6217279 on Build GRCh38
rs34280304 Chr.11: 6217429-6217429 on Build GRCh38
rs760623418 Chr.11: 6217299-6217299 on Build GRCh38
rs575583059 Chr.11: 6217432-6217432 on Build GRCh38

View Full Product Details