Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49131242-49131489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC3A
Entrez Gene ID
22862
Gene Name
fibronectin type III domain containing 3A
Gene Aliases
FNDC3, HUGO, bA203I16.1, bA203I16.5
UniGene
Hs.508010
Gene Chromosome Location
Chr.13: 48975241-49209779 on Build GRCh38
Transcripts
NM_001079673.1 NM_001278438.1 NM_014923.4 NR_103528.1 XM_011534997.2 XM_017020440.1 XM_017020441.1 XM_017020442.1 XM_017020443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047942 c.389C>T p.P130L Chr.13: 49131273-49131273 on Build GRCh38
4957006 c.461A>G p.Y154C Chr.13: 49131345-49131345 on Build GRCh38
4975775 c.394C>A p.P132T Chr.13: 49131278-49131278 on Build GRCh38
1942519 c.373_374insT p.T127fs*49 Chr.13: 49131257-49131258 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202097950 Chr.13: 49131337-49131337 on Build GRCh38
rs751635737 Chr.13: 49131284-49131284 on Build GRCh38
rs763226463 Chr.13: 49131365-49131365 on Build GRCh38
rs778604916 Chr.13: 49131321-49131321 on Build GRCh38
rs760964832 Chr.13: 49131414-49131414 on Build GRCh38
rs753801214 Chr.13: 49131423-49131424 on Build GRCh38
rs192490696 Chr.13: 49131404-49131404 on Build GRCh38
rs746784986 Chr.13: 49131343-49131343 on Build GRCh38
rs931713333 Chr.13: 49131416-49131416 on Build GRCh38
rs774465353 Chr.13: 49131394-49131394 on Build GRCh38
rs944631158 Chr.13: 49131448-49131448 on Build GRCh38
rs368383893 Chr.13: 49131278-49131278 on Build GRCh38
rs368752642 Chr.13: 49131422-49131422 on Build GRCh38
rs775713516 Chr.13: 49131274-49131274 on Build GRCh38
rs199834112 Chr.13: 49131302-49131302 on Build GRCh38
rs551407649 Chr.13: 49131320-49131320 on Build GRCh38
rs767989864 Chr.13: 49131289-49131289 on Build GRCh38
rs537141481 Chr.13: 49131435-49131435 on Build GRCh38
rs779581907 Chr.13: 49131299-49131299 on Build GRCh38
rs764214776 Chr.13: 49131280-49131280 on Build GRCh38
rs548270596 Chr.13: 49131398-49131398 on Build GRCh38
rs200997716 Chr.13: 49131391-49131391 on Build GRCh38
rs376740097 Chr.13: 49131363-49131363 on Build GRCh38
rs371847209 Chr.13: 49131348-49131348 on Build GRCh38
rs770701778 Chr.13: 49131354-49131354 on Build GRCh38
rs758844482 Chr.13: 49131421-49131421 on Build GRCh38
rs765409328 Chr.13: 49131273-49131273 on Build GRCh38
rs762348761 Chr.13: 49131288-49131288 on Build GRCh38
rs760466037 Chr.13: 49131266-49131266 on Build GRCh38
rs375105881 Chr.13: 49131406-49131406 on Build GRCh38
rs750775139 Chr.13: 49131298-49131298 on Build GRCh38
rs761833142 Chr.13: 49131396-49131396 on Build GRCh38
rs772153929 Chr.13: 49131340-49131340 on Build GRCh38
rs777722532 Chr.13: 49131342-49131342 on Build GRCh38
rs1036201161 Chr.13: 49131460-49131460 on Build GRCh38
rs774638514 Chr.13: 49131301-49131301 on Build GRCh38

View Full Product Details