Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7487240-7487481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163
Entrez Gene ID
9332
Gene Name
CD163 molecule
Gene Aliases
M130, MM130, SCARI1
UniGene
Hs.504641
Gene Chromosome Location
Chr.12: 7470816-7503818 on Build GRCh38
Transcripts
NM_004244.5 NM_203416.3 XM_005253528.4 XM_005253529.4 XM_017020231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6519943 c.1970A>T p.Q657L Chr.12: 7487439-7487439 on Build GRCh38
3688447 c.2105C>T p.A702V Chr.12: 7487403-7487403 on Build GRCh38
4862782 c.2088T>C p.P696P Chr.12: 7487420-7487420 on Build GRCh38
4914864 c.1937G>T p.W646L Chr.12: 7487472-7487472 on Build GRCh38
416064 c.2023G>A p.E675K Chr.12: 7487386-7487386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976890411 Chr.12: 7487439-7487439 on Build GRCh38
rs778668873 Chr.12: 7487373-7487373 on Build GRCh38
rs905332718 Chr.12: 7487300-7487300 on Build GRCh38
rs746479491 Chr.12: 7487421-7487421 on Build GRCh38
rs759825589 Chr.12: 7487336-7487336 on Build GRCh38
rs745589563 Chr.12: 7487383-7487383 on Build GRCh38
rs753684494 Chr.12: 7487354-7487354 on Build GRCh38
rs770235496 Chr.12: 7487434-7487434 on Build GRCh38
rs923227343 Chr.12: 7487361-7487361 on Build GRCh38
rs763338760 Chr.12: 7487309-7487309 on Build GRCh38
rs201457023 Chr.12: 7487346-7487346 on Build GRCh38
rs752761728 Chr.12: 7487344-7487344 on Build GRCh38
rs779555857 Chr.12: 7487413-7487413 on Build GRCh38
rs188505613 Chr.12: 7487285-7487285 on Build GRCh38
rs149501156 Chr.12: 7487440-7487440 on Build GRCh38
rs767709351 Chr.12: 7487340-7487340 on Build GRCh38
rs774035317 Chr.12: 7487453-7487453 on Build GRCh38
rs1015262274 Chr.12: 7487422-7487422 on Build GRCh38
rs983070305 Chr.12: 7487419-7487419 on Build GRCh38
rs962055706 Chr.12: 7487327-7487327 on Build GRCh38
rs369519795 Chr.12: 7487345-7487345 on Build GRCh38
rs953497216 Chr.12: 7487368-7487368 on Build GRCh38
rs868312071 Chr.12: 7487392-7487392 on Build GRCh38
rs757129074 Chr.12: 7487359-7487359 on Build GRCh38
rs763355747 Chr.12: 7487449-7487449 on Build GRCh38
rs751888043 Chr.12: 7487328-7487328 on Build GRCh38
rs150335436 Chr.12: 7487310-7487310 on Build GRCh38
rs965071363 Chr.12: 7487431-7487431 on Build GRCh38
rs771647321 Chr.12: 7487390-7487390 on Build GRCh38

View Full Product Details