Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41539639-41539912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS4
Entrez Gene ID
84296
Gene Name
GINS complex subunit 4
Gene Aliases
SLD5
UniGene
Hs.656996
Gene Chromosome Location
Chr.8: 41529206-41545046 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Gene Symbol
LOC102723729
Entrez Gene ID
102723729
Gene Name
uncharacterized LOC102723729
Gene Aliases
-
UniGene
Hs.586384
Gene Chromosome Location
Chr.8: 41534305-41578368 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1658895 c.371C>G p.P124R Chr.8: 41539751-41539751 on Build GRCh38
3942660 c.299T>C p.I100T Chr.8: 41539679-41539679 on Build GRCh38
3925297 c.352G>A p.E118K Chr.8: 41539732-41539732 on Build GRCh38
3699088 c.317A>G p.H106R Chr.8: 41539697-41539697 on Build GRCh38
1253242 c.306G>C p.K102N Chr.8: 41539686-41539686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748867438 Chr.8: 41539795-41539795 on Build GRCh38
rs746836266 Chr.8: 41539670-41539670 on Build GRCh38
rs376006720 Chr.8: 41539801-41539801 on Build GRCh38
rs773987379 Chr.8: 41539774-41539777 on Build GRCh38
rs199761157 Chr.8: 41539709-41539709 on Build GRCh38
rs754751330 Chr.8: 41539761-41539761 on Build GRCh38
rs200745887 Chr.8: 41539732-41539732 on Build GRCh38
rs765312724 Chr.8: 41539865-41539865 on Build GRCh38
rs1043880257 Chr.8: 41539860-41539860 on Build GRCh38
rs554016887 Chr.8: 41539863-41539863 on Build GRCh38
rs113767927 Chr.8: 41539782-41539782 on Build GRCh38
rs941369356 Chr.8: 41539764-41539764 on Build GRCh38
rs763468665 Chr.8: 41539728-41539728 on Build GRCh38
rs751494503 Chr.8: 41539760-41539760 on Build GRCh38
rs74703449 Chr.8: 41539735-41539735 on Build GRCh38
rs373246125 Chr.8: 41539869-41539869 on Build GRCh38
rs764466719 Chr.8: 41539820-41539820 on Build GRCh38
rs754878809 Chr.8: 41539873-41539873 on Build GRCh38
rs999237925 Chr.8: 41539697-41539697 on Build GRCh38
rs767898710 Chr.8: 41539736-41539736 on Build GRCh38
rs770394312 Chr.8: 41539796-41539796 on Build GRCh38
rs983501163 Chr.8: 41539845-41539845 on Build GRCh38
rs768474635 Chr.8: 41539711-41539712 on Build GRCh38
rs760990539 Chr.8: 41539810-41539810 on Build GRCh38
rs775623850 Chr.8: 41539797-41539797 on Build GRCh38
rs768468703 Chr.8: 41539684-41539684 on Build GRCh38
rs771487958 Chr.8: 41539729-41539729 on Build GRCh38
rs559432599 Chr.8: 41539846-41539846 on Build GRCh38
rs752880102 Chr.8: 41539739-41539739 on Build GRCh38
rs752549914 Chr.8: 41539876-41539876 on Build GRCh38
rs777393250 Chr.8: 41539792-41539792 on Build GRCh38
rs779897589 Chr.8: 41539759-41539759 on Build GRCh38
rs775856426 Chr.8: 41539806-41539806 on Build GRCh38
rs951532434 Chr.8: 41539715-41539715 on Build GRCh38
rs757398107 Chr.8: 41539757-41539757 on Build GRCh38
rs774531389 Chr.8: 41539852-41539852 on Build GRCh38
rs772351026 Chr.8: 41539802-41539802 on Build GRCh38
rs778923827 Chr.8: 41539749-41539749 on Build GRCh38
rs1031148596 Chr.8: 41539727-41539727 on Build GRCh38
rs780758152 Chr.8: 41539685-41539685 on Build GRCh38
rs892297364 Chr.8: 41539842-41539842 on Build GRCh38
rs550157200 Chr.8: 41539712-41539712 on Build GRCh38
rs771978178 Chr.8: 41539790-41539790 on Build GRCh38
rs372725593 Chr.8: 41539791-41539791 on Build GRCh38
rs372132452 Chr.8: 41539714-41539714 on Build GRCh38
rs141057295 Chr.8: 41539683-41539683 on Build GRCh38
rs142553733 Chr.8: 41539752-41539752 on Build GRCh38
rs760938896 Chr.8: 41539743-41539743 on Build GRCh38
rs942103430 Chr.8: 41539868-41539868 on Build GRCh38
rs376381703 Chr.8: 41539751-41539751 on Build GRCh38
rs767232488 Chr.8: 41539874-41539874 on Build GRCh38
rs755900572 Chr.8: 41539877-41539877 on Build GRCh38
rs747787378 Chr.8: 41539698-41539698 on Build GRCh38

View Full Product Details