Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87313744-87314017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B13
Entrez Gene ID
345275
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
HMFN0376, NIIL497, SCDR9, SDR16C3
UniGene
Hs.284414
Gene Chromosome Location
Chr.4: 87303789-87322906 on Build GRCh38
Transcripts
NM_001136230.2 NM_178135.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3606822 c.673T>C p.F225L Chr.4: 87313845-87313845 on Build GRCh38
1694643 c.590G>A p.R197K Chr.4: 87313928-87313928 on Build GRCh38
3606823 c.615C>T p.A205A Chr.4: 87313903-87313903 on Build GRCh38
3606824 c.602C>T p.S201L Chr.4: 87313916-87313916 on Build GRCh38
7086785 c.590G>C p.R197T Chr.4: 87313928-87313928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552091804 Chr.4: 87313832-87313832 on Build GRCh38
rs754693984 Chr.4: 87313853-87313853 on Build GRCh38
rs760737421 Chr.4: 87313954-87313954 on Build GRCh38
rs772092560 Chr.4: 87313875-87313875 on Build GRCh38
rs750477403 Chr.4: 87313919-87313919 on Build GRCh38
rs775983018 Chr.4: 87313774-87313774 on Build GRCh38
rs372241178 Chr.4: 87313893-87313893 on Build GRCh38
rs762314183 Chr.4: 87313928-87313928 on Build GRCh38
rs371697119 Chr.4: 87313842-87313842 on Build GRCh38
rs372756309 Chr.4: 87313846-87313846 on Build GRCh38
rs866695150 Chr.4: 87313946-87313946 on Build GRCh38
rs559107427 Chr.4: 87313773-87313773 on Build GRCh38
rs778958020 Chr.4: 87313862-87313862 on Build GRCh38
rs768927517 Chr.4: 87313922-87313922 on Build GRCh38
rs368298699 Chr.4: 87313972-87313972 on Build GRCh38
rs530612782 Chr.4: 87313783-87313783 on Build GRCh38
rs376004820 Chr.4: 87313905-87313905 on Build GRCh38
rs774741984 Chr.4: 87313924-87313924 on Build GRCh38
rs755178310 Chr.4: 87313971-87313971 on Build GRCh38
rs1048748638 Chr.4: 87313835-87313835 on Build GRCh38
rs780123324 Chr.4: 87313836-87313836 on Build GRCh38
rs80182459 Chr.4: 87313945-87313945 on Build GRCh38
rs748058364 Chr.4: 87313868-87313868 on Build GRCh38
rs142648167 Chr.4: 87313916-87313916 on Build GRCh38
rs537764715 Chr.4: 87313977-87313977 on Build GRCh38
rs752472433 Chr.4: 87313989-87313989 on Build GRCh38
rs552212851 Chr.4: 87313942-87313942 on Build GRCh38
rs372499266 Chr.4: 87313995-87313995 on Build GRCh38
rs988233524 Chr.4: 87313811-87313811 on Build GRCh38
rs751990890 Chr.4: 87313793-87313793 on Build GRCh38
rs993184958 Chr.4: 87313833-87313833 on Build GRCh38
rs201455716 Chr.4: 87313870-87313870 on Build GRCh38
rs1018465850 Chr.4: 87313934-87313934 on Build GRCh38
rs756162852 Chr.4: 87313945-87313945 on Build GRCh38
rs867111910 Chr.4: 87313894-87313894 on Build GRCh38
rs954775837 Chr.4: 87313864-87313864 on Build GRCh38
rs766659694 Chr.4: 87313961-87313961 on Build GRCh38
rs750439655 Chr.4: 87313829-87313829 on Build GRCh38
rs897529199 Chr.4: 87313809-87313809 on Build GRCh38
rs547498403 Chr.4: 87313944-87313944 on Build GRCh38
rs763222231 Chr.4: 87313780-87313780 on Build GRCh38
rs762262933 Chr.4: 87313797-87313797 on Build GRCh38
rs566033418 Chr.4: 87313967-87313967 on Build GRCh38
rs570425723 Chr.4: 87313889-87313889 on Build GRCh38
rs773686851 Chr.4: 87313857-87313857 on Build GRCh38
rs777425194 Chr.4: 87313887-87313887 on Build GRCh38
rs767617317 Chr.4: 87313802-87313802 on Build GRCh38

View Full Product Details