Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48462259-48462500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48455707-48466110 on Build GRCh38
Transcripts
NM_013348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211524 c.538G>A p.V180I Chr.19: 48462262-48462262 on Build GRCh38
6413665 c.670C>T p.R224C Chr.19: 48462394-48462394 on Build GRCh38
5549062 c.621G>T p.A207A Chr.19: 48462345-48462345 on Build GRCh38
6374692 c.562A>G p.M188V Chr.19: 48462286-48462286 on Build GRCh38
4656309 c.626G>A p.R209H Chr.19: 48462350-48462350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772889207 Chr.19: 48462417-48462417 on Build GRCh38
rs761242323 Chr.19: 48462302-48462302 on Build GRCh38
rs758513522 Chr.19: 48462370-48462370 on Build GRCh38
rs765441291 Chr.19: 48462464-48462464 on Build GRCh38
rs774201345 Chr.19: 48462444-48462444 on Build GRCh38
rs202116675 Chr.19: 48462353-48462353 on Build GRCh38
rs575240795 Chr.19: 48462443-48462443 on Build GRCh38
rs747712026 Chr.19: 48462409-48462409 on Build GRCh38
rs780492911 Chr.19: 48462378-48462378 on Build GRCh38
rs762697691 Chr.19: 48462419-48462419 on Build GRCh38
rs779330593 Chr.19: 48462364-48462364 on Build GRCh38
rs776127890 Chr.19: 48462278-48462278 on Build GRCh38
rs1047058267 Chr.19: 48462434-48462434 on Build GRCh38
rs781660850 Chr.19: 48462406-48462406 on Build GRCh38
rs369764191 Chr.19: 48462345-48462345 on Build GRCh38
rs769206780 Chr.19: 48462383-48462383 on Build GRCh38
rs376747285 Chr.19: 48462315-48462315 on Build GRCh38
rs770440037 Chr.19: 48462421-48462421 on Build GRCh38
rs764934165 Chr.19: 48462311-48462311 on Build GRCh38
rs766166537 Chr.19: 48462319-48462319 on Build GRCh38
rs1003086708 Chr.19: 48462451-48462451 on Build GRCh38
rs759246543 Chr.19: 48462447-48462447 on Build GRCh38
rs762783258 Chr.19: 48462317-48462317 on Build GRCh38
rs759532571 Chr.19: 48462342-48462342 on Build GRCh38
rs754040372 Chr.19: 48462347-48462347 on Build GRCh38
rs992466355 Chr.19: 48462463-48462463 on Build GRCh38
rs756364785 Chr.19: 48462456-48462456 on Build GRCh38
rs751142404 Chr.19: 48462472-48462472 on Build GRCh38
rs898977413 Chr.19: 48462468-48462468 on Build GRCh38
rs757400397 Chr.19: 48462348-48462348 on Build GRCh38
rs769269839 Chr.19: 48462412-48462412 on Build GRCh38
rs35733575 Chr.19: 48462310-48462310 on Build GRCh38
rs767291622 Chr.19: 48462344-48462344 on Build GRCh38
rs1011345315 Chr.19: 48462392-48462392 on Build GRCh38
rs762026869 Chr.19: 48462459-48462459 on Build GRCh38
rs556659563 Chr.19: 48462366-48462366 on Build GRCh38
rs767621898 Chr.19: 48462454-48462454 on Build GRCh38
rs538564251 Chr.19: 48462336-48462336 on Build GRCh38
rs747275683 Chr.19: 48462382-48462382 on Build GRCh38
rs370442458 Chr.19: 48462388-48462388 on Build GRCh38
rs772836127 Chr.19: 48462316-48462316 on Build GRCh38
rs1044555417 Chr.19: 48462424-48462424 on Build GRCh38
rs934245512 Chr.19: 48462423-48462423 on Build GRCh38

View Full Product Details