Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40366918-40367181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DISP2
Entrez Gene ID
85455
Gene Name
dispatched RND transporter family member 2
Gene Aliases
DISPB, HsT16908, LINC00594
UniGene
Hs.355645
Gene Chromosome Location
Chr.15: 40354359-40378755 on Build GRCh38
Transcripts
NM_033510.2 XM_011522125.2 XM_017022687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2269345 c.1049G>A p.R350H Chr.15: 40367161-40367161 on Build GRCh38
1708011 c.1047C>A p.N349K Chr.15: 40367159-40367159 on Build GRCh38
4054334 c.1002C>T p.C334C Chr.15: 40367114-40367114 on Build GRCh38
5797355 c.1063G>T p.D355Y Chr.15: 40367175-40367175 on Build GRCh38
6838747 c.1011C>G p.S337R Chr.15: 40367123-40367123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921337988 Chr.15: 40367016-40367016 on Build GRCh38
rs200137211 Chr.15: 40367065-40367065 on Build GRCh38
rs569973086 Chr.15: 40366999-40366999 on Build GRCh38
rs759415920 Chr.15: 40367079-40367079 on Build GRCh38
rs779431494 Chr.15: 40367140-40367140 on Build GRCh38
rs753931363 Chr.15: 40367120-40367120 on Build GRCh38
rs756145205 Chr.15: 40367113-40367113 on Build GRCh38
rs776391352 Chr.15: 40367045-40367045 on Build GRCh38
rs908522335 Chr.15: 40366985-40366985 on Build GRCh38
rs954343650 Chr.15: 40367036-40367036 on Build GRCh38
rs768158565 Chr.15: 40367018-40367018 on Build GRCh38
rs950428447 Chr.15: 40367135-40367135 on Build GRCh38
rs143054067 Chr.15: 40367062-40367062 on Build GRCh38
rs746816637 Chr.15: 40366960-40366960 on Build GRCh38
rs369585136 Chr.15: 40367161-40367161 on Build GRCh38
rs902477711 Chr.15: 40366952-40366952 on Build GRCh38
rs747594647 Chr.15: 40367047-40367047 on Build GRCh38
rs368084105 Chr.15: 40367061-40367061 on Build GRCh38
rs778371849 Chr.15: 40367083-40367083 on Build GRCh38
rs1003191504 Chr.15: 40367114-40367114 on Build GRCh38
rs746270882 Chr.15: 40367009-40367009 on Build GRCh38
rs535155561 Chr.15: 40367100-40367100 on Build GRCh38
rs1007000591 Chr.15: 40367044-40367044 on Build GRCh38
rs746192268 Chr.15: 40367141-40367141 on Build GRCh38
rs769492125 Chr.15: 40367049-40367049 on Build GRCh38
rs757716771 Chr.15: 40367130-40367130 on Build GRCh38
rs998230382 Chr.15: 40366966-40366966 on Build GRCh38
rs201498467 Chr.15: 40367095-40367095 on Build GRCh38
rs34475634 Chr.15: 40367078-40367078 on Build GRCh38
rs919022822 Chr.15: 40367090-40367090 on Build GRCh38
rs983004205 Chr.15: 40366971-40366971 on Build GRCh38
rs756551657 Chr.15: 40367027-40367027 on Build GRCh38
rs1033379595 Chr.15: 40367155-40367155 on Build GRCh38
rs779192987 Chr.15: 40367007-40367007 on Build GRCh38
rs758851870 Chr.15: 40367160-40367160 on Build GRCh38

View Full Product Details