Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206961100-206961325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCAMR
Entrez Gene ID
83953
Gene Name
Fc fragment of IgA and IgM receptor
Gene Aliases
CD351, FCA/MR, FKSG87
UniGene
Hs.145519
Gene Chromosome Location
Chr.1: 206957955-206970657 on Build GRCh38
Transcripts
NM_001122979.2 NM_001170631.1 NM_032029.4 XM_011510051.2 XM_011510052.2 XM_017002442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4505078 c.683C>T p.P228L Chr.1: 206961193-206961193 on Build GRCh38
3803701 c.633G>C p.M211I Chr.1: 206961171-206961171 on Build GRCh38
3803700 c.705G>C p.M235I Chr.1: 206961171-206961171 on Build GRCh38
4558669 c.703G>T p.G235W Chr.1: 206961101-206961101 on Build GRCh38
5426395 c.581-2A>G p.? Chr.1: 206961225-206961225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751883557 Chr.1: 206961204-206961204 on Build GRCh38
rs970275554 Chr.1: 206961250-206961250 on Build GRCh38
rs200011175 Chr.1: 206961169-206961169 on Build GRCh38
rs1041108255 Chr.1: 206961143-206961143 on Build GRCh38
rs1032024961 Chr.1: 206961188-206961188 on Build GRCh38
rs768213670 Chr.1: 206961182-206961182 on Build GRCh38
rs893573373 Chr.1: 206961211-206961211 on Build GRCh38
rs763503910 Chr.1: 206961174-206961174 on Build GRCh38
rs901277909 Chr.1: 206961170-206961170 on Build GRCh38
rs376426554 Chr.1: 206961185-206961185 on Build GRCh38
rs944004647 Chr.1: 206961125-206961125 on Build GRCh38
rs117620733 Chr.1: 206961246-206961246 on Build GRCh38
rs954402599 Chr.1: 206961206-206961206 on Build GRCh38
rs757629990 Chr.1: 206961218-206961218 on Build GRCh38
rs747912949 Chr.1: 206961120-206961120 on Build GRCh38
rs201258954 Chr.1: 206961208-206961208 on Build GRCh38
rs556798122 Chr.1: 206961138-206961138 on Build GRCh38
rs541351315 Chr.1: 206961273-206961273 on Build GRCh38
rs369380685 Chr.1: 206961157-206961157 on Build GRCh38
rs889695963 Chr.1: 206961124-206961124 on Build GRCh38
rs771122836 Chr.1: 206961133-206961133 on Build GRCh38
rs552971213 Chr.1: 206961219-206961219 on Build GRCh38
rs372580744 Chr.1: 206961203-206961203 on Build GRCh38
rs1038601734 Chr.1: 206961181-206961182 on Build GRCh38
rs373309798 Chr.1: 206961153-206961153 on Build GRCh38
rs977074432 Chr.1: 206961268-206961268 on Build GRCh38

View Full Product Details