Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 78029034-78029307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC3A
Entrez Gene ID
10143
Gene Name
C-type lectin domain family 3 member A
Gene Aliases
CLECSF1
UniGene
Hs.177936
Gene Chromosome Location
Chr.16: 78022546-78032106 on Build GRCh38
Transcripts
NM_001244755.1 NM_005752.4

SNP Detail

SNP ID Chromosome Location
rs767197237 Chr.16: 78029180-78029180 on Build GRCh38
rs150644547 Chr.16: 78029237-78029237 on Build GRCh38
rs1029568760 Chr.16: 78029259-78029259 on Build GRCh38
rs75678536 Chr.16: 78029267-78029267 on Build GRCh38
rs7192416 Chr.16: 78029246-78029246 on Build GRCh38
rs966223836 Chr.16: 78029083-78029083 on Build GRCh38
rs1036685062 Chr.16: 78029275-78029275 on Build GRCh38
rs758894612 Chr.16: 78029195-78029195 on Build GRCh38
rs141712944 Chr.16: 78029069-78029069 on Build GRCh38
rs7190832 Chr.16: 78029167-78029167 on Build GRCh38
rs931690759 Chr.16: 78029127-78029127 on Build GRCh38
rs940615179 Chr.16: 78029165-78029165 on Build GRCh38
rs886698072 Chr.16: 78029099-78029099 on Build GRCh38
rs895237425 Chr.16: 78029224-78029224 on Build GRCh38
rs547405831 Chr.16: 78029109-78029109 on Build GRCh38
rs940973462 Chr.16: 78029271-78029271 on Build GRCh38
rs1031225893 Chr.16: 78029247-78029247 on Build GRCh38
rs544685539 Chr.16: 78029245-78029245 on Build GRCh38
rs187111111 Chr.16: 78029230-78029230 on Build GRCh38
rs908875734 Chr.16: 78029159-78029159 on Build GRCh38
rs7197560 Chr.16: 78029103-78029103 on Build GRCh38
rs966670874 Chr.16: 78029253-78029253 on Build GRCh38
rs898234069 Chr.16: 78029137-78029137 on Build GRCh38
rs992996166 Chr.16: 78029265-78029265 on Build GRCh38
rs976286292 Chr.16: 78029257-78029257 on Build GRCh38
rs907092863 Chr.16: 78029175-78029175 on Build GRCh38
rs927374568 Chr.16: 78029252-78029252 on Build GRCh38
rs1048793901 Chr.16: 78029142-78029142 on Build GRCh38
rs779607776 Chr.16: 78029179-78029179 on Build GRCh38
rs903845064 Chr.16: 78029241-78029241 on Build GRCh38

View Full Product Details