Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104949438-104949675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP4
Entrez Gene ID
837
Gene Name
caspase 4
Gene Aliases
ICE(rel)II, ICEREL-II, ICH-2, Mih1, Mih1/TX, TX
UniGene
Hs.138378
Gene Chromosome Location
Chr.11: 104942866-104968598 on Build GRCh38
Transcripts
NM_001225.3 NM_033306.2 XM_011543019.1 XM_017018397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
21797 c.757A>G p.I253V Chr.11: 104949567-104949567 on Build GRCh38
5837684 c.734G>A p.S245N Chr.11: 104949590-104949590 on Build GRCh38
922426 c.754G>A p.V252I Chr.11: 104949570-104949570 on Build GRCh38
7105706 c.679G>A p.D227N Chr.11: 104949645-104949645 on Build GRCh38
305557 c.722G>A p.R241H Chr.11: 104949602-104949602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755130733 Chr.11: 104949639-104949639 on Build GRCh38
rs560386687 Chr.11: 104949492-104949492 on Build GRCh38
rs1012753929 Chr.11: 104949540-104949540 on Build GRCh38
rs370828951 Chr.11: 104949533-104949533 on Build GRCh38
rs748269121 Chr.11: 104949497-104949497 on Build GRCh38
rs752966299 Chr.11: 104949646-104949646 on Build GRCh38
rs200458703 Chr.11: 104949656-104949656 on Build GRCh38
rs150393983 Chr.11: 104949643-104949643 on Build GRCh38
rs140509533 Chr.11: 104949559-104949559 on Build GRCh38
rs148312912 Chr.11: 104949557-104949557 on Build GRCh38
rs138280041 Chr.11: 104949648-104949648 on Build GRCh38
rs1035479466 Chr.11: 104949521-104949521 on Build GRCh38
rs906608658 Chr.11: 104949484-104949484 on Build GRCh38
rs368813530 Chr.11: 104949603-104949603 on Build GRCh38
rs72486010 Chr.11: 104949481-104949481 on Build GRCh38
rs968674919 Chr.11: 104949583-104949583 on Build GRCh38
rs755292245 Chr.11: 104949487-104949487 on Build GRCh38
rs766388990 Chr.11: 104949616-104949616 on Build GRCh38
rs756530960 Chr.11: 104949524-104949524 on Build GRCh38
rs1020749906 Chr.11: 104949648-104949649 on Build GRCh38
rs374502870 Chr.11: 104949574-104949574 on Build GRCh38
rs35170829 Chr.11: 104949573-104949574 on Build GRCh38
rs1046195364 Chr.11: 104949530-104949530 on Build GRCh38
rs201303104 Chr.11: 104949604-104949604 on Build GRCh38
rs768110001 Chr.11: 104949600-104949600 on Build GRCh38
rs751739258 Chr.11: 104949636-104949636 on Build GRCh38
rs775043878 Chr.11: 104949562-104949562 on Build GRCh38
rs776381340 Chr.11: 104949602-104949602 on Build GRCh38
rs775854988 Chr.11: 104949656-104949658 on Build GRCh38
rs746488096 Chr.11: 104949563-104949563 on Build GRCh38
rs372801724 Chr.11: 104949614-104949614 on Build GRCh38
rs749646670 Chr.11: 104949655-104949655 on Build GRCh38
rs905135501 Chr.11: 104949570-104949570 on Build GRCh38
rs760855980 Chr.11: 104949626-104949626 on Build GRCh38

View Full Product Details