Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135572696-135572969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS6L
Entrez Gene ID
203522
Gene Name
integrator complex subunit 6 like
Gene Aliases
DDX26B
UniGene
Hs.496829
Gene Chromosome Location
Chr.X: 135520630-135582535 on Build GRCh38
Transcripts
NM_182540.4 XM_005262394.2 XM_006724740.2 XM_006724741.3 XM_017029342.1 XM_017029343.1 XM_017029344.1 XM_017029345.1 XM_017029346.1

SNP Detail

SNP ID Chromosome Location
rs782399791 Chr.X: 135572903-135572903 on Build GRCh38
rs781914825 Chr.X: 135572894-135572894 on Build GRCh38
rs782504690 Chr.X: 135572847-135572847 on Build GRCh38
rs78975820 Chr.X: 135572737-135572737 on Build GRCh38
rs782471664 Chr.X: 135572801-135572801 on Build GRCh38
rs781793840 Chr.X: 135572837-135572837 on Build GRCh38
rs782261612 Chr.X: 135572846-135572846 on Build GRCh38
rs782726535 Chr.X: 135572728-135572728 on Build GRCh38
rs782654510 Chr.X: 135572842-135572842 on Build GRCh38
rs782685748 Chr.X: 135572778-135572778 on Build GRCh38
rs782771777 Chr.X: 135572797-135572797 on Build GRCh38
rs781849806 Chr.X: 135572798-135572798 on Build GRCh38
rs373280783 Chr.X: 135572817-135572817 on Build GRCh38
rs782024008 Chr.X: 135572788-135572788 on Build GRCh38
rs782108311 Chr.X: 135572940-135572940 on Build GRCh38
rs782139524 Chr.X: 135572790-135572790 on Build GRCh38
rs61732230 Chr.X: 135572922-135572922 on Build GRCh38
rs782566417 Chr.X: 135572839-135572839 on Build GRCh38
rs377640430 Chr.X: 135572863-135572863 on Build GRCh38
rs782209316 Chr.X: 135572865-135572865 on Build GRCh38
rs373384691 Chr.X: 135572726-135572726 on Build GRCh38
rs782314200 Chr.X: 135572873-135572873 on Build GRCh38
rs374389347 Chr.X: 135572918-135572918 on Build GRCh38
rs371386844 Chr.X: 135572902-135572902 on Build GRCh38

View Full Product Details