Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30379697-30379944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPD
Entrez Gene ID
1362
Gene Name
carboxypeptidase D
Gene Aliases
GP180
UniGene
Hs.446079
Gene Chromosome Location
Chr.17: 30378924-30469657 on Build GRCh38
Transcripts
NM_001304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5740936 c.737G>A p.R246H Chr.17: 30379717-30379717 on Build GRCh38
5767760 c.725T>C p.I242T Chr.17: 30379705-30379705 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029417772 Chr.17: 30379817-30379817 on Build GRCh38
rs989760481 Chr.17: 30379897-30379897 on Build GRCh38
rs746141912 Chr.17: 30379756-30379756 on Build GRCh38
rs754596617 Chr.17: 30379732-30379732 on Build GRCh38
rs777880754 Chr.17: 30379745-30379745 on Build GRCh38
rs764724603 Chr.17: 30379733-30379733 on Build GRCh38
rs534532633 Chr.17: 30379752-30379752 on Build GRCh38
rs113652936 Chr.17: 30379917-30379917 on Build GRCh38
rs758447620 Chr.17: 30379741-30379741 on Build GRCh38
rs1017877119 Chr.17: 30379880-30379880 on Build GRCh38
rs779657259 Chr.17: 30379756-30379757 on Build GRCh38
rs111266957 Chr.17: 30379796-30379796 on Build GRCh38
rs538241703 Chr.17: 30379911-30379911 on Build GRCh38
rs775186988 Chr.17: 30379841-30379841 on Build GRCh38
rs770042036 Chr.17: 30379766-30379766 on Build GRCh38
rs974614819 Chr.17: 30379807-30379807 on Build GRCh38
rs774739325 Chr.17: 30379715-30379715 on Build GRCh38
rs62068698 Chr.17: 30379898-30379898 on Build GRCh38
rs187036801 Chr.17: 30379809-30379809 on Build GRCh38
rs545895021 Chr.17: 30379768-30379768 on Build GRCh38
rs1006545791 Chr.17: 30379870-30379870 on Build GRCh38
rs986509026 Chr.17: 30379847-30379847 on Build GRCh38
rs912211230 Chr.17: 30379860-30379860 on Build GRCh38
rs890874659 Chr.17: 30379832-30379832 on Build GRCh38
rs1055120512 Chr.17: 30379751-30379751 on Build GRCh38
rs749336733 Chr.17: 30379771-30379771 on Build GRCh38
rs767874233 Chr.17: 30379718-30379718 on Build GRCh38
rs757460095 Chr.17: 30379888-30379888 on Build GRCh38
rs751489515 Chr.17: 30379750-30379750 on Build GRCh38
rs768683244 Chr.17: 30379776-30379776 on Build GRCh38
rs966905031 Chr.17: 30379903-30379903 on Build GRCh38
rs201196265 Chr.17: 30379716-30379716 on Build GRCh38
rs567150494 Chr.17: 30379721-30379721 on Build GRCh38
rs752243799 Chr.17: 30379738-30379738 on Build GRCh38
rs762197393 Chr.17: 30379717-30379717 on Build GRCh38
rs1022934409 Chr.17: 30379901-30379901 on Build GRCh38
rs1035424108 Chr.17: 30379739-30379739 on Build GRCh38

View Full Product Details