Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136668486-136668745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL7
Entrez Gene ID
51162
Gene Name
EGF like domain multiple 7
Gene Aliases
NEU1, VE-STATIN, ZNEU1
UniGene
Hs.91481
Gene Chromosome Location
Chr.9: 136654753-136672678 on Build GRCh38
Transcripts
NM_016215.4 NM_201446.2 NR_045110.1 NR_045111.1 NR_046367.1 XM_006717141.3 XM_011518764.1 XM_011518765.1 XM_011518766.1 XM_011518767.1 XM_011518768.1 XM_017014795.1 XM_017014796.1 XM_017014797.1 XM_017014798.1 XM_017014799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6931859 c.101G>A p.R34Q Chr.9: 136668577-136668577 on Build GRCh38
6944333 c.167G>A p.C56Y Chr.9: 136668643-136668643 on Build GRCh38
4588634 c.106C>T p.H36Y Chr.9: 136668582-136668582 on Build GRCh38
7365942 c.133G>C p.V45L Chr.9: 136668609-136668609 on Build GRCh38
239682 c.113A>T p.D38V Chr.9: 136668589-136668589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758520402 Chr.9: 136668576-136668576 on Build GRCh38
rs548014889 Chr.9: 136668559-136668559 on Build GRCh38
rs113610188 Chr.9: 136668529-136668529 on Build GRCh38
rs150164841 Chr.9: 136668661-136668661 on Build GRCh38
rs770792123 Chr.9: 136668673-136668673 on Build GRCh38
rs2297534 Chr.9: 136668639-136668639 on Build GRCh38
rs754217396 Chr.9: 136668721-136668721 on Build GRCh38
rs916872498 Chr.9: 136668590-136668590 on Build GRCh38
rs760679475 Chr.9: 136668564-136668564 on Build GRCh38
rs765213699 Chr.9: 136668507-136668507 on Build GRCh38
rs781632992 Chr.9: 136668533-136668533 on Build GRCh38
rs746395482 Chr.9: 136668655-136668656 on Build GRCh38
rs998201166 Chr.9: 136668651-136668651 on Build GRCh38
rs368404574 Chr.9: 136668628-136668628 on Build GRCh38
rs752653098 Chr.9: 136668573-136668573 on Build GRCh38
rs747784641 Chr.9: 136668608-136668608 on Build GRCh38
rs748165103 Chr.9: 136668557-136668557 on Build GRCh38
rs374275228 Chr.9: 136668616-136668616 on Build GRCh38
rs1019299438 Chr.9: 136668680-136668680 on Build GRCh38
rs150932253 Chr.9: 136668605-136668605 on Build GRCh38
rs780611350 Chr.9: 136668596-136668596 on Build GRCh38
rs940127360 Chr.9: 136668629-136668629 on Build GRCh38
rs566214627 Chr.9: 136668531-136668531 on Build GRCh38
rs1006591968 Chr.9: 136668538-136668538 on Build GRCh38
rs768123998 Chr.9: 136668553-136668553 on Build GRCh38
rs568237914 Chr.9: 136668584-136668584 on Build GRCh38
rs750663449 Chr.9: 136668706-136668706 on Build GRCh38
rs764175431 Chr.9: 136668582-136668582 on Build GRCh38
rs371572647 Chr.9: 136668645-136668645 on Build GRCh38
rs781270000 Chr.9: 136668542-136668542 on Build GRCh38
rs767392646 Chr.9: 136668558-136668558 on Build GRCh38
rs766745501 Chr.9: 136668716-136668716 on Build GRCh38
rs770381358 Chr.9: 136668680-136668703 on Build GRCh38
rs761882012 Chr.9: 136668695-136668695 on Build GRCh38
rs888551652 Chr.9: 136668521-136668521 on Build GRCh38
rs2297535 Chr.9: 136668692-136668692 on Build GRCh38
rs756512511 Chr.9: 136668644-136668644 on Build GRCh38
rs114919674 Chr.9: 136668693-136668693 on Build GRCh38
rs766407929 Chr.9: 136668572-136668572 on Build GRCh38
rs770717845 Chr.9: 136668615-136668615 on Build GRCh38
rs759493745 Chr.9: 136668688-136668688 on Build GRCh38
rs143961691 Chr.9: 136668600-136668600 on Build GRCh38
rs770381056 Chr.9: 136668537-136668537 on Build GRCh38
rs757508218 Chr.9: 136668585-136668585 on Build GRCh38
rs371810712 Chr.9: 136668611-136668611 on Build GRCh38
rs140748161 Chr.9: 136668655-136668655 on Build GRCh38
rs760905571 Chr.9: 136668713-136668713 on Build GRCh38
rs746734078 Chr.9: 136668672-136668672 on Build GRCh38
rs747167688 Chr.9: 136668703-136668703 on Build GRCh38
rs375443368 Chr.9: 136668522-136668522 on Build GRCh38
rs750714980 Chr.9: 136668641-136668641 on Build GRCh38
rs755672766 Chr.9: 136668683-136668683 on Build GRCh38
rs747317686 Chr.9: 136668528-136668528 on Build GRCh38
rs776329468 Chr.9: 136668676-136668676 on Build GRCh38
rs746343047 Chr.9: 136668535-136668535 on Build GRCh38
rs764201416 Chr.9: 136668622-136668622 on Build GRCh38
rs192259203 Chr.9: 136668527-136668527 on Build GRCh38
rs755489905 Chr.9: 136668654-136668654 on Build GRCh38
rs757350195 Chr.9: 136668533-136668534 on Build GRCh38
rs754341907 Chr.9: 136668647-136668647 on Build GRCh38
rs4880118 Chr.9: 136668541-136668541 on Build GRCh38
rs762008786 Chr.9: 136668638-136668638 on Build GRCh38
rs200663721 Chr.9: 136668604-136668604 on Build GRCh38
rs755508641 Chr.9: 136668722-136668722 on Build GRCh38
rs781514618 Chr.9: 136668588-136668588 on Build GRCh38
rs762900850 Chr.9: 136668620-136668620 on Build GRCh38
rs747731431 Chr.9: 136668657-136668657 on Build GRCh38
rs771912424 Chr.9: 136668609-136668609 on Build GRCh38
rs771297117 Chr.9: 136668724-136668724 on Build GRCh38
rs200785608 Chr.9: 136668552-136668552 on Build GRCh38
rs199679849 Chr.9: 136668591-136668591 on Build GRCh38
rs950942092 Chr.9: 136668577-136668577 on Build GRCh38
rs577218094 Chr.9: 136668674-136668674 on Build GRCh38
rs369211581 Chr.9: 136668698-136668698 on Build GRCh38
rs777582115 Chr.9: 136668667-136668667 on Build GRCh38
rs752880505 Chr.9: 136668519-136668519 on Build GRCh38

View Full Product Details