Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132887222-132887475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA9
Entrez Gene ID
11092
Gene Name
sperm acrosome associated 9
Gene Aliases
C9orf9, Mast
UniGene
Hs.62595
Gene Chromosome Location
Chr.9: 132878320-132890314 on Build GRCh38
Transcripts
NM_001316897.1 NM_001316898.1 NM_001316900.1 NM_018956.4 NR_133631.1 XM_017014231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905164 c.188G>A p.R63Q Chr.9: 132887412-132887412 on Build GRCh38
4885959 c.177C>A p.S59R Chr.9: 132887401-132887401 on Build GRCh38
4371687 c.196C>T p.L66L Chr.9: 132887420-132887420 on Build GRCh38
3905169 c.199C>T p.L67F Chr.9: 132887423-132887423 on Build GRCh38
4885960 c.177C>A p.S59R Chr.9: 132887401-132887401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367975616 Chr.9: 132887337-132887337 on Build GRCh38
rs750333522 Chr.9: 132887452-132887452 on Build GRCh38
rs746737674 Chr.9: 132887301-132887302 on Build GRCh38
rs772769293 Chr.9: 132887422-132887422 on Build GRCh38
rs774754661 Chr.9: 132887340-132887340 on Build GRCh38
rs563367778 Chr.9: 132887426-132887426 on Build GRCh38
rs769502830 Chr.9: 132887329-132887329 on Build GRCh38
rs186145280 Chr.9: 132887355-132887355 on Build GRCh38
rs780624092 Chr.9: 132887324-132887324 on Build GRCh38
rs748604453 Chr.9: 132887418-132887418 on Build GRCh38
rs779726103 Chr.9: 132887410-132887410 on Build GRCh38
rs774270483 Chr.9: 132887437-132887437 on Build GRCh38
rs751472684 Chr.9: 132887388-132887388 on Build GRCh38
rs748642379 Chr.9: 132887343-132887343 on Build GRCh38
rs559224424 Chr.9: 132887310-132887310 on Build GRCh38
rs555310723 Chr.9: 132887259-132887259 on Build GRCh38
rs898034756 Chr.9: 132887279-132887279 on Build GRCh38
rs776970822 Chr.9: 132887358-132887358 on Build GRCh38
rs999764867 Chr.9: 132887284-132887284 on Build GRCh38
rs374237797 Chr.9: 132887379-132887379 on Build GRCh38
rs766762257 Chr.9: 132887357-132887357 on Build GRCh38
rs763802525 Chr.9: 132887433-132887433 on Build GRCh38
rs929476612 Chr.9: 132887307-132887307 on Build GRCh38
rs375286305 Chr.9: 132887335-132887335 on Build GRCh38
rs755599580 Chr.9: 132887409-132887409 on Build GRCh38
rs199653353 Chr.9: 132887412-132887412 on Build GRCh38
rs755656682 Chr.9: 132887454-132887454 on Build GRCh38
rs915548367 Chr.9: 132887269-132887269 on Build GRCh38
rs777022236 Chr.9: 132887430-132887430 on Build GRCh38
rs140770799 Chr.9: 132887276-132887276 on Build GRCh38
rs759752984 Chr.9: 132887431-132887431 on Build GRCh38
rs772054797 Chr.9: 132887347-132887347 on Build GRCh38
rs773357134 Chr.9: 132887354-132887354 on Build GRCh38
rs779276098 Chr.9: 132887416-132887416 on Build GRCh38
rs201660249 Chr.9: 132887415-132887415 on Build GRCh38
rs749993742 Chr.9: 132887400-132887400 on Build GRCh38
rs200219145 Chr.9: 132887327-132887327 on Build GRCh38
rs767439490 Chr.9: 132887399-132887399 on Build GRCh38
rs759044522 Chr.9: 132887271-132887271 on Build GRCh38
rs201066708 Chr.9: 132887446-132887446 on Build GRCh38
rs754577385 Chr.9: 132887397-132887397 on Build GRCh38
rs929841556 Chr.9: 132887374-132887374 on Build GRCh38
rs918421026 Chr.9: 132887361-132887361 on Build GRCh38
rs762696204 Chr.9: 132887359-132887359 on Build GRCh38
rs769038978 Chr.9: 132887338-132887338 on Build GRCh38
rs143097194 Chr.9: 132887425-132887425 on Build GRCh38
rs77377515 Chr.9: 132887250-132887250 on Build GRCh38
rs960071070 Chr.9: 132887424-132887424 on Build GRCh38
rs11243925 Chr.9: 132887309-132887309 on Build GRCh38
rs150690462 Chr.9: 132887438-132887438 on Build GRCh38
rs890549192 Chr.9: 132887325-132887325 on Build GRCh38

View Full Product Details