Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207468458-207468683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR2
Entrez Gene ID
1380
Gene Name
complement component 3d receptor 2
Gene Aliases
C3DR, CD21, CR, CVID7, SLEB9
UniGene
Hs.445757
Gene Chromosome Location
Chr.1: 207454300-207489895 on Build GRCh38
Transcripts
NM_001006658.2 NM_001877.4 XM_011509206.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482633 c.480C>T p.I160I Chr.1: 207468561-207468561 on Build GRCh38
3482632 c.477G>A p.M159I Chr.1: 207468558-207468558 on Build GRCh38
7362600 c.524C>T p.P175L Chr.1: 207468605-207468605 on Build GRCh38
3482631 c.472C>T p.P158S Chr.1: 207468553-207468553 on Build GRCh38
5907868 c.599C>T p.S200L Chr.1: 207468680-207468680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758596478 Chr.1: 207468506-207468506 on Build GRCh38
rs539720446 Chr.1: 207468538-207468538 on Build GRCh38
rs75282758 Chr.1: 207468605-207468605 on Build GRCh38
rs369202942 Chr.1: 207468542-207468542 on Build GRCh38
rs138760137 Chr.1: 207468533-207468533 on Build GRCh38
rs751783758 Chr.1: 207468596-207468596 on Build GRCh38
rs778616639 Chr.1: 207468645-207468645 on Build GRCh38
rs201223793 Chr.1: 207468649-207468649 on Build GRCh38
rs186581459 Chr.1: 207468487-207468487 on Build GRCh38
rs911878039 Chr.1: 207468503-207468503 on Build GRCh38
rs746314457 Chr.1: 207468528-207468528 on Build GRCh38
rs533297833 Chr.1: 207468537-207468537 on Build GRCh38
rs780000327 Chr.1: 207468498-207468499 on Build GRCh38
rs764271765 Chr.1: 207468508-207468508 on Build GRCh38
rs755365484 Chr.1: 207468638-207468638 on Build GRCh38
rs768228152 Chr.1: 207468486-207468486 on Build GRCh38
rs757345336 Chr.1: 207468514-207468514 on Build GRCh38
rs1054517610 Chr.1: 207468572-207468572 on Build GRCh38
rs190990627 Chr.1: 207468566-207468566 on Build GRCh38
rs760648455 Chr.1: 207468568-207468568 on Build GRCh38
rs778471118 Chr.1: 207468549-207468549 on Build GRCh38
rs750347465 Chr.1: 207468614-207468614 on Build GRCh38
rs936251537 Chr.1: 207468550-207468550 on Build GRCh38
rs961288010 Chr.1: 207468488-207468488 on Build GRCh38
rs776543969 Chr.1: 207468574-207468574 on Build GRCh38
rs761278296 Chr.1: 207468491-207468491 on Build GRCh38
rs766207448 Chr.1: 207468499-207468499 on Build GRCh38
rs764369159 Chr.1: 207468595-207468595 on Build GRCh38
rs780591393 Chr.1: 207468617-207468617 on Build GRCh38
rs200800206 Chr.1: 207468608-207468608 on Build GRCh38
rs751623731 Chr.1: 207468513-207468513 on Build GRCh38
rs759285664 Chr.1: 207468588-207468588 on Build GRCh38
rs377004857 Chr.1: 207468500-207468500 on Build GRCh38
rs985682133 Chr.1: 207468515-207468515 on Build GRCh38
rs2063143 Chr.1: 207468495-207468495 on Build GRCh38
rs771013353 Chr.1: 207468569-207468569 on Build GRCh38
rs373598320 Chr.1: 207468496-207468496 on Build GRCh38
rs747691020 Chr.1: 207468556-207468556 on Build GRCh38
rs757046729 Chr.1: 207468535-207468535 on Build GRCh38
rs972622640 Chr.1: 207468490-207468490 on Build GRCh38
rs781713410 Chr.1: 207468518-207468518 on Build GRCh38
rs771541633 Chr.1: 207468560-207468560 on Build GRCh38
rs370389616 Chr.1: 207468601-207468601 on Build GRCh38
rs751006774 Chr.1: 207468526-207468527 on Build GRCh38
rs373493698 Chr.1: 207468629-207468629 on Build GRCh38
rs754171470 Chr.1: 207468620-207468620 on Build GRCh38

View Full Product Details