Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 103120672-103120899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
UniGene
Hs.503721
Gene Chromosome Location
Chr.11: 103109431-103479863 on Build GRCh38
Transcripts
NM_001080463.1 NM_001377.2 XM_006718903.2 XM_017018291.1 XM_017018292.1 XM_017018293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6053539 c.1246C>A p.Q416K Chr.11: 103120800-103120800 on Build GRCh38
398665 c.1238G>A p.S413N Chr.11: 103120792-103120792 on Build GRCh38
1603958 c.1139T>C p.L380S Chr.11: 103120693-103120693 on Build GRCh38
922339 c.1213A>C p.N405H Chr.11: 103120767-103120767 on Build GRCh38
5804147 c.1247A>T p.Q416L Chr.11: 103120801-103120801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200822573 Chr.11: 103120738-103120738 on Build GRCh38
rs529456510 Chr.11: 103120860-103120860 on Build GRCh38
rs777978053 Chr.11: 103120732-103120732 on Build GRCh38
rs1054885310 Chr.11: 103120862-103120862 on Build GRCh38
rs973779500 Chr.11: 103120824-103120824 on Build GRCh38
rs752575739 Chr.11: 103120851-103120851 on Build GRCh38
rs116888435 Chr.11: 103120787-103120787 on Build GRCh38
rs747358182 Chr.11: 103120785-103120785 on Build GRCh38
rs541038013 Chr.11: 103120711-103120711 on Build GRCh38
rs772351140 Chr.11: 103120767-103120767 on Build GRCh38
rs893511644 Chr.11: 103120867-103120867 on Build GRCh38
rs765929492 Chr.11: 103120813-103120813 on Build GRCh38
rs777277500 Chr.11: 103120797-103120797 on Build GRCh38
rs202130799 Chr.11: 103120810-103120810 on Build GRCh38
rs1035807816 Chr.11: 103120860-103120861 on Build GRCh38
rs563587072 Chr.11: 103120853-103120853 on Build GRCh38
rs534849587 Chr.11: 103120826-103120826 on Build GRCh38
rs1005893204 Chr.11: 103120838-103120838 on Build GRCh38
rs762610855 Chr.11: 103120812-103120812 on Build GRCh38
rs746033439 Chr.11: 103120756-103120756 on Build GRCh38
rs779012837 Chr.11: 103120744-103120744 on Build GRCh38
rs369614706 Chr.11: 103120705-103120705 on Build GRCh38
rs1015494874 Chr.11: 103120846-103120847 on Build GRCh38
rs991557585 Chr.11: 103120869-103120869 on Build GRCh38
rs773881152 Chr.11: 103120816-103120816 on Build GRCh38
rs928128188 Chr.11: 103120858-103120858 on Build GRCh38
rs775878881 Chr.11: 103120778-103120778 on Build GRCh38
rs767054315 Chr.11: 103120840-103120840 on Build GRCh38
rs373327850 Chr.11: 103120739-103120739 on Build GRCh38

View Full Product Details