Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19444204-19444477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf39
Entrez Gene ID
128977
Gene Name
chromosome 22 open reading frame 39
Gene Aliases
-
UniGene
Hs.659565
Gene Chromosome Location
Chr.22: 19440886-19448232 on Build GRCh38
Transcripts
NM_001166242.1 NM_173793.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1032098 c.233C>A p.A78E Chr.22: 19444350-19444350 on Build GRCh38
1032099 c.215G>A p.R72Q Chr.22: 19444368-19444368 on Build GRCh38
2892277 c.221G>A p.R74Q Chr.22: 19444362-19444362 on Build GRCh38
4667547 c.423C>T p.D141D Chr.22: 19444271-19444271 on Build GRCh38
3708134 c.193C>A p.Q65K Chr.22: 19444390-19444390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375560562 Chr.22: 19444356-19444356 on Build GRCh38
rs766777566 Chr.22: 19444391-19444391 on Build GRCh38
rs375609529 Chr.22: 19444343-19444343 on Build GRCh38
rs766548902 Chr.22: 19444347-19444347 on Build GRCh38
rs773360630 Chr.22: 19444232-19444232 on Build GRCh38
rs774176418 Chr.22: 19444381-19444381 on Build GRCh38
rs1021039040 Chr.22: 19444243-19444243 on Build GRCh38
rs1042606950 Chr.22: 19444455-19444455 on Build GRCh38
rs770903685 Chr.22: 19444307-19444308 on Build GRCh38
rs762288546 Chr.22: 19444337-19444337 on Build GRCh38
rs902105197 Chr.22: 19444447-19444447 on Build GRCh38
rs1056219854 Chr.22: 19444401-19444401 on Build GRCh38
rs765198676 Chr.22: 19444237-19444237 on Build GRCh38
rs373255364 Chr.22: 19444363-19444363 on Build GRCh38
rs144484392 Chr.22: 19444348-19444348 on Build GRCh38
rs774844853 Chr.22: 19444313-19444313 on Build GRCh38
rs762769936 Chr.22: 19444425-19444426 on Build GRCh38
rs767953102 Chr.22: 19444339-19444339 on Build GRCh38
rs776821639 Chr.22: 19444403-19444404 on Build GRCh38
rs768390938 Chr.22: 19444322-19444322 on Build GRCh38
rs768231083 Chr.22: 19444380-19444380 on Build GRCh38
rs368195480 Chr.22: 19444261-19444261 on Build GRCh38
rs61748355 Chr.22: 19444271-19444271 on Build GRCh38
rs766314898 Chr.22: 19444262-19444262 on Build GRCh38
rs748784717 Chr.22: 19444373-19444373 on Build GRCh38
rs959656634 Chr.22: 19444325-19444325 on Build GRCh38
rs556903035 Chr.22: 19444453-19444453 on Build GRCh38
rs761285765 Chr.22: 19444390-19444390 on Build GRCh38
rs755997492 Chr.22: 19444369-19444369 on Build GRCh38
rs777588773 Chr.22: 19444274-19444274 on Build GRCh38
rs369983280 Chr.22: 19444368-19444368 on Build GRCh38
rs773783525 Chr.22: 19444340-19444340 on Build GRCh38
rs570937669 Chr.22: 19444308-19444308 on Build GRCh38
rs780790474 Chr.22: 19444367-19444367 on Build GRCh38
rs747746008 Chr.22: 19444383-19444383 on Build GRCh38
rs754147776 Chr.22: 19444270-19444270 on Build GRCh38
rs765469124 Chr.22: 19444357-19444357 on Build GRCh38
rs537072711 Chr.22: 19444388-19444388 on Build GRCh38
rs990809198 Chr.22: 19444326-19444326 on Build GRCh38
rs973949401 Chr.22: 19444324-19444324 on Build GRCh38
rs746788815 Chr.22: 19444277-19444277 on Build GRCh38
rs745722844 Chr.22: 19444285-19444285 on Build GRCh38
rs56867241 Chr.22: 19444313-19444313 on Build GRCh38
rs373031322 Chr.22: 19444427-19444427 on Build GRCh38
rs942373678 Chr.22: 19444402-19444402 on Build GRCh38
rs749032612 Chr.22: 19444319-19444319 on Build GRCh38
rs777151337 Chr.22: 19444394-19444394 on Build GRCh38
rs535158179 Chr.22: 19444331-19444331 on Build GRCh38
rs578011205 Chr.22: 19444372-19444372 on Build GRCh38
rs113884318 Chr.22: 19444456-19444456 on Build GRCh38
rs200451846 Chr.22: 19444421-19444421 on Build GRCh38
rs756999666 Chr.22: 19444279-19444279 on Build GRCh38
rs368988639 Chr.22: 19444289-19444289 on Build GRCh38
rs149705263 Chr.22: 19444282-19444282 on Build GRCh38
rs763145981 Chr.22: 19444436-19444436 on Build GRCh38
rs192542043 Chr.22: 19444249-19444249 on Build GRCh38
rs752878945 Chr.22: 19444362-19444362 on Build GRCh38
rs1040654294 Chr.22: 19444441-19444441 on Build GRCh38
rs997646711 Chr.22: 19444236-19444236 on Build GRCh38

View Full Product Details