Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34152300-34152533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
UniGene
Hs.23361
Gene Chromosome Location
Chr.11: 34150987-34358008 on Build GRCh38
Transcripts
NM_145804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197857 c.3044G>A p.R1015H Chr.11: 34152421-34152421 on Build GRCh38
139885 c.2453C>T p.P818L Chr.11: 34152454-34152454 on Build GRCh38
926618 c.2513G>A p.R838Q Chr.11: 34152394-34152394 on Build GRCh38
1604502 c.3069C>A p.S1023S Chr.11: 34152396-34152396 on Build GRCh38
6368406 c.2381A>T p.K794M Chr.11: 34152526-34152526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747667990 Chr.11: 34152506-34152506 on Build GRCh38
rs777083770 Chr.11: 34152337-34152354 on Build GRCh38
rs369160367 Chr.11: 34152367-34152367 on Build GRCh38
rs764266917 Chr.11: 34152473-34152473 on Build GRCh38
rs139428758 Chr.11: 34152376-34152376 on Build GRCh38
rs776143668 Chr.11: 34152422-34152422 on Build GRCh38
rs749551117 Chr.11: 34152411-34152411 on Build GRCh38
rs760298525 Chr.11: 34152355-34152355 on Build GRCh38
rs761492145 Chr.11: 34152440-34152440 on Build GRCh38
rs148487808 Chr.11: 34152417-34152417 on Build GRCh38
rs1052295344 Chr.11: 34152394-34152394 on Build GRCh38
rs139861196 Chr.11: 34152438-34152438 on Build GRCh38
rs747581476 Chr.11: 34152497-34152497 on Build GRCh38
rs372778906 Chr.11: 34152404-34152404 on Build GRCh38
rs757656341 Chr.11: 34152415-34152415 on Build GRCh38
rs764914019 Chr.11: 34152446-34152446 on Build GRCh38
rs141021753 Chr.11: 34152499-34152499 on Build GRCh38
rs144645011 Chr.11: 34152361-34152361 on Build GRCh38
rs765555625 Chr.11: 34152440-34152441 on Build GRCh38
rs557470544 Chr.11: 34152377-34152377 on Build GRCh38
rs1020919510 Chr.11: 34152340-34152340 on Build GRCh38
rs756226554 Chr.11: 34152395-34152395 on Build GRCh38
rs1009557274 Chr.11: 34152332-34152332 on Build GRCh38
rs746443994 Chr.11: 34152427-34152427 on Build GRCh38
rs199813409 Chr.11: 34152500-34152500 on Build GRCh38
rs146433580 Chr.11: 34152435-34152435 on Build GRCh38
rs143317941 Chr.11: 34152462-34152462 on Build GRCh38
rs150248775 Chr.11: 34152509-34152509 on Build GRCh38
rs1044983094 Chr.11: 34152326-34152326 on Build GRCh38
rs866649017 Chr.11: 34152387-34152387 on Build GRCh38
rs555615662 Chr.11: 34152328-34152328 on Build GRCh38
rs144583805 Chr.11: 34152390-34152390 on Build GRCh38
rs757533385 Chr.11: 34152479-34152479 on Build GRCh38
rs116440254 Chr.11: 34152420-34152420 on Build GRCh38
rs746167793 Chr.11: 34152482-34152482 on Build GRCh38
rs756102220 Chr.11: 34152471-34152471 on Build GRCh38
rs114304923 Chr.11: 34152485-34152485 on Build GRCh38
rs368398114 Chr.11: 34152459-34152459 on Build GRCh38
rs376690299 Chr.11: 34152366-34152366 on Build GRCh38
rs750255959 Chr.11: 34152374-34152374 on Build GRCh38
rs371133755 Chr.11: 34152481-34152481 on Build GRCh38
rs968064972 Chr.11: 34152341-34152341 on Build GRCh38
rs200312319 Chr.11: 34152486-34152486 on Build GRCh38
rs374914955 Chr.11: 34152452-34152452 on Build GRCh38
rs759655917 Chr.11: 34152378-34152378 on Build GRCh38
rs377195382 Chr.11: 34152421-34152421 on Build GRCh38
rs941832281 Chr.11: 34152337-34152337 on Build GRCh38

View Full Product Details