Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90020854-90021117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR1
Entrez Gene ID
1666
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
Gene Aliases
DECR, NADPH, SDR18C1
UniGene
Hs.492212
Gene Chromosome Location
Chr.8: 90001337-90052004 on Build GRCh38
Transcripts
NM_001359.1 XM_005250808.4 XM_011516880.2 XM_017013147.1 XM_017013148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686581 c.418-2A>G p.? Chr.8: 90020907-90020907 on Build GRCh38
7364042 c.550A>T p.I184F Chr.8: 90021041-90021041 on Build GRCh38
6472730 c.457A>T p.T153S Chr.8: 90020948-90020948 on Build GRCh38
7247956 c.522C>T p.F174F Chr.8: 90021013-90021013 on Build GRCh38
270755 c.470C>T p.S157F Chr.8: 90020961-90020961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946424372 Chr.8: 90020974-90020974 on Build GRCh38
rs752317734 Chr.8: 90020913-90020913 on Build GRCh38
rs755032279 Chr.8: 90020956-90020956 on Build GRCh38
rs752880164 Chr.8: 90021083-90021083 on Build GRCh38
rs781289544 Chr.8: 90020965-90020965 on Build GRCh38
rs758826963 Chr.8: 90020937-90020937 on Build GRCh38
rs756182345 Chr.8: 90021091-90021091 on Build GRCh38
rs769944318 Chr.8: 90020987-90020987 on Build GRCh38
rs766798793 Chr.8: 90021060-90021060 on Build GRCh38
rs763902500 Chr.8: 90021034-90021034 on Build GRCh38
rs774205872 Chr.8: 90021009-90021009 on Build GRCh38
rs778011226 Chr.8: 90021095-90021095 on Build GRCh38
rs142348077 Chr.8: 90020990-90020990 on Build GRCh38
rs752035746 Chr.8: 90021062-90021062 on Build GRCh38
rs752998702 Chr.8: 90021049-90021049 on Build GRCh38
rs747525935 Chr.8: 90020951-90020951 on Build GRCh38
rs774268008 Chr.8: 90020893-90020893 on Build GRCh38
rs763457044 Chr.8: 90021058-90021058 on Build GRCh38
rs913654862 Chr.8: 90020994-90020994 on Build GRCh38
rs375650574 Chr.8: 90021077-90021077 on Build GRCh38
rs137904244 Chr.8: 90020923-90020923 on Build GRCh38
rs190568496 Chr.8: 90021012-90021012 on Build GRCh38
rs766361708 Chr.8: 90020892-90020892 on Build GRCh38
rs780743047 Chr.8: 90020944-90020944 on Build GRCh38
rs765704982 Chr.8: 90020921-90020921 on Build GRCh38
rs989205910 Chr.8: 90020980-90020980 on Build GRCh38
rs760262701 Chr.8: 90021021-90021021 on Build GRCh38
rs748322078 Chr.8: 90020975-90020975 on Build GRCh38
rs372630127 Chr.8: 90021014-90021014 on Build GRCh38
rs201468153 Chr.8: 90021057-90021057 on Build GRCh38
rs759009481 Chr.8: 90020899-90020899 on Build GRCh38
rs370070181 Chr.8: 90021078-90021078 on Build GRCh38
rs770682603 Chr.8: 90021000-90021000 on Build GRCh38
rs981028164 Chr.8: 90020915-90020915 on Build GRCh38
rs777957430 Chr.8: 90020991-90020991 on Build GRCh38
rs756597062 Chr.8: 90020907-90020907 on Build GRCh38
rs1052014064 Chr.8: 90020922-90020922 on Build GRCh38
rs753506368 Chr.8: 90021050-90021050 on Build GRCh38
rs755755254 Chr.8: 90020916-90020916 on Build GRCh38
rs759924542 Chr.8: 90020909-90020909 on Build GRCh38
rs932550108 Chr.8: 90021080-90021080 on Build GRCh38
rs143170054 Chr.8: 90021013-90021013 on Build GRCh38

View Full Product Details