Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46574276-46574503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
UniGene
Hs.129955
Gene Chromosome Location
Chr.X: 46573687-46598496 on Build GRCh38
Transcripts
NM_019886.3

SNP Detail

SNP ID Chromosome Location
rs773697310 Chr.X: 46574445-46574445 on Build GRCh38
rs777553106 Chr.X: 46574369-46574369 on Build GRCh38
rs746614653 Chr.X: 46574420-46574420 on Build GRCh38
rs778970910 Chr.X: 46574402-46574402 on Build GRCh38
rs774424444 Chr.X: 46574483-46574483 on Build GRCh38
rs745409653 Chr.X: 46574407-46574407 on Build GRCh38
rs181984333 Chr.X: 46574464-46574464 on Build GRCh38
rs749031627 Chr.X: 46574372-46574372 on Build GRCh38
rs376196221 Chr.X: 46574387-46574387 on Build GRCh38
rs766923926 Chr.X: 46574452-46574452 on Build GRCh38
rs371271901 Chr.X: 46574484-46574484 on Build GRCh38
rs752864505 Chr.X: 46574350-46574350 on Build GRCh38
rs755870456 Chr.X: 46574365-46574365 on Build GRCh38
rs764208188 Chr.X: 46574482-46574482 on Build GRCh38
rs752917425 Chr.X: 46574476-46574476 on Build GRCh38
rs373483256 Chr.X: 46574319-46574319 on Build GRCh38
rs770059621 Chr.X: 46574439-46574439 on Build GRCh38
rs956147595 Chr.X: 46574340-46574340 on Build GRCh38
rs200081268 Chr.X: 46574310-46574310 on Build GRCh38
rs774811144 Chr.X: 46574453-46574453 on Build GRCh38
rs61734022 Chr.X: 46574447-46574447 on Build GRCh38
rs921002483 Chr.X: 46574459-46574459 on Build GRCh38
rs761449271 Chr.X: 46574335-46574335 on Build GRCh38
rs767525052 Chr.X: 46574468-46574468 on Build GRCh38
rs771669492 Chr.X: 46574408-46574408 on Build GRCh38
rs751698930 Chr.X: 46574315-46574315 on Build GRCh38
rs761020258 Chr.X: 46574477-46574477 on Build GRCh38

View Full Product Details