Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78214262-78214495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387453 c.972G>A p.P324P Chr.17: 78214420-78214420 on Build GRCh38
7357525 c.25G>T p.A9S Chr.17: 78214341-78214341 on Build GRCh38
1716121 c.58T>A p.S20T Chr.17: 78214374-78214374 on Build GRCh38
5101109 c.1000G>A p.A334T Chr.17: 78214392-78214392 on Build GRCh38
1386406 c.76C>T p.P26S Chr.17: 78214392-78214392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567991073 Chr.17: 78214475-78214475 on Build GRCh38
rs768778329 Chr.17: 78214476-78214476 on Build GRCh38
rs370361101 Chr.17: 78214400-78214400 on Build GRCh38
rs372753451 Chr.17: 78214375-78214375 on Build GRCh38
rs752405432 Chr.17: 78214287-78214287 on Build GRCh38
rs899470863 Chr.17: 78214439-78214439 on Build GRCh38
rs9904341 Chr.17: 78214286-78214286 on Build GRCh38
rs751292269 Chr.17: 78214414-78214414 on Build GRCh38
rs373880047 Chr.17: 78214307-78214307 on Build GRCh38
rs763927641 Chr.17: 78214290-78214290 on Build GRCh38
rs749745235 Chr.17: 78214310-78214310 on Build GRCh38
rs770250176 Chr.17: 78214306-78214306 on Build GRCh38
rs556970411 Chr.17: 78214449-78214450 on Build GRCh38
rs760898261 Chr.17: 78214474-78214474 on Build GRCh38
rs757193907 Chr.17: 78214292-78214292 on Build GRCh38
rs754589913 Chr.17: 78214417-78214417 on Build GRCh38
rs543367853 Chr.17: 78214339-78214339 on Build GRCh38
rs753843703 Chr.17: 78214291-78214291 on Build GRCh38
rs750450342 Chr.17: 78214304-78214305 on Build GRCh38
rs748438531 Chr.17: 78214297-78214297 on Build GRCh38
rs148424824 Chr.17: 78214421-78214421 on Build GRCh38
rs759731481 Chr.17: 78214330-78214330 on Build GRCh38
rs202101234 Chr.17: 78214293-78214293 on Build GRCh38
rs527915990 Chr.17: 78214440-78214440 on Build GRCh38
rs775441897 Chr.17: 78214470-78214470 on Build GRCh38
rs143160639 Chr.17: 78214341-78214341 on Build GRCh38
rs779792860 Chr.17: 78214296-78214296 on Build GRCh38
rs771587762 Chr.17: 78214314-78214314 on Build GRCh38
rs199661984 Chr.17: 78214460-78214460 on Build GRCh38
rs998676813 Chr.17: 78214451-78214451 on Build GRCh38
rs982941178 Chr.17: 78214352-78214352 on Build GRCh38
rs145526644 Chr.17: 78214406-78214406 on Build GRCh38
rs532219684 Chr.17: 78214358-78214358 on Build GRCh38
rs765188612 Chr.17: 78214393-78214393 on Build GRCh38
rs920106823 Chr.17: 78214338-78214338 on Build GRCh38
rs771536267 Chr.17: 78214430-78214430 on Build GRCh38
rs745932468 Chr.17: 78214459-78214459 on Build GRCh38
rs774532555 Chr.17: 78214320-78214320 on Build GRCh38
rs779557679 Chr.17: 78214410-78214410 on Build GRCh38
rs762112729 Chr.17: 78214285-78214285 on Build GRCh38
rs1022618408 Chr.17: 78214445-78214445 on Build GRCh38
rs1043005222 Chr.17: 78214442-78214442 on Build GRCh38
rs969829480 Chr.17: 78214453-78214453 on Build GRCh38
rs267605071 Chr.17: 78214392-78214392 on Build GRCh38
rs755988912 Chr.17: 78214289-78214289 on Build GRCh38

View Full Product Details