Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10761984-10762209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTR9
Entrez Gene ID
9646
Gene Name
CTR9 homolog, Paf1/RNA polymerase II complex component
Gene Aliases
SH2BP1, TSBP, p150, p150TSP
UniGene
Hs.725151
Gene Chromosome Location
Chr.11: 10750987-10779755 on Build GRCh38
Transcripts
NM_014633.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389543 c.827C>A p.A276E Chr.11: 10762032-10762032 on Build GRCh38
1604027 c.815T>G p.L272W Chr.11: 10762020-10762020 on Build GRCh38
1627746 c.849+4G>A p.? Chr.11: 10762058-10762058 on Build GRCh38
2110106 c.800G>A p.S267N Chr.11: 10762005-10762005 on Build GRCh38
7197527 c.786_787insT p.T263fs*3 Chr.11: 10761991-10761992 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035220551 Chr.11: 10762156-10762156 on Build GRCh38
rs773751641 Chr.11: 10762104-10762104 on Build GRCh38
rs894316929 Chr.11: 10762142-10762142 on Build GRCh38
rs375558859 Chr.11: 10762079-10762079 on Build GRCh38
rs1040240276 Chr.11: 10762050-10762050 on Build GRCh38
rs1016150075 Chr.11: 10762172-10762172 on Build GRCh38
rs768620591 Chr.11: 10762096-10762096 on Build GRCh38
rs893112874 Chr.11: 10762061-10762061 on Build GRCh38
rs901340474 Chr.11: 10762062-10762062 on Build GRCh38
rs963294561 Chr.11: 10762180-10762180 on Build GRCh38
rs755592846 Chr.11: 10762018-10762018 on Build GRCh38
rs1039995559 Chr.11: 10762058-10762058 on Build GRCh38
rs770400083 Chr.11: 10762103-10762103 on Build GRCh38
rs772611433 Chr.11: 10762082-10762082 on Build GRCh38
rs745636253 Chr.11: 10762086-10762086 on Build GRCh38
rs576086623 Chr.11: 10762089-10762089 on Build GRCh38
rs566772038 Chr.11: 10762090-10762090 on Build GRCh38
rs1035897025 Chr.11: 10762115-10762115 on Build GRCh38
rs1012764019 Chr.11: 10762151-10762151 on Build GRCh38
rs372407563 Chr.11: 10762075-10762075 on Build GRCh38
rs201745405 Chr.11: 10762017-10762017 on Build GRCh38
rs774743339 Chr.11: 10762107-10762107 on Build GRCh38
rs969506809 Chr.11: 10762120-10762120 on Build GRCh38
rs1016621131 Chr.11: 10762106-10762106 on Build GRCh38
rs763594613 Chr.11: 10762024-10762024 on Build GRCh38
rs767044434 Chr.11: 10762011-10762011 on Build GRCh38
rs747191546 Chr.11: 10762092-10762092 on Build GRCh38
rs34459041 Chr.11: 10762055-10762055 on Build GRCh38

View Full Product Details