Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1022128-1022399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGRN
Entrez Gene ID
375790
Gene Name
agrin
Gene Aliases
CMS8, CMSPPD
UniGene
Hs.273330
Gene Chromosome Location
Chr.1: 1020102-1056119 on Build GRCh38
Transcripts
NM_001305275.1 NM_198576.3 XM_005244749.3 XM_011541429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4634475 c.370C>A p.L124M Chr.1: 1022369-1022369 on Build GRCh38
4577666 c.384C>G p.H128Q Chr.1: 1022383-1022383 on Build GRCh38
6596436 c.273G>A p.K91K Chr.1: 1022272-1022272 on Build GRCh38
3806182 c.275T>G p.V92G Chr.1: 1022274-1022274 on Build GRCh38
4637004 c.205C>T p.R69W Chr.1: 1022204-1022204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907583629 Chr.1: 1022197-1022197 on Build GRCh38
rs373066821 Chr.1: 1022360-1022360 on Build GRCh38
rs746863220 Chr.1: 1022162-1022162 on Build GRCh38
rs756623659 Chr.1: 1022225-1022225 on Build GRCh38
rs750748055 Chr.1: 1022224-1022224 on Build GRCh38
rs928130162 Chr.1: 1022168-1022168 on Build GRCh38
rs769659786 Chr.1: 1022263-1022263 on Build GRCh38
rs773486550 Chr.1: 1022297-1022297 on Build GRCh38
rs776504646 Chr.1: 1022185-1022185 on Build GRCh38
rs760057776 Chr.1: 1022355-1022355 on Build GRCh38
rs142684737 Chr.1: 1022213-1022213 on Build GRCh38
rs368144442 Chr.1: 1022244-1022244 on Build GRCh38
rs781159362 Chr.1: 1022261-1022261 on Build GRCh38
rs755949709 Chr.1: 1022379-1022379 on Build GRCh38
rs768159179 Chr.1: 1022269-1022269 on Build GRCh38
rs6657048 Chr.1: 1022260-1022260 on Build GRCh38
rs755100104 Chr.1: 1022250-1022250 on Build GRCh38
rs754384811 Chr.1: 1022346-1022346 on Build GRCh38
rs138696597 Chr.1: 1022362-1022362 on Build GRCh38
rs761093589 Chr.1: 1022305-1022305 on Build GRCh38
rs762519724 Chr.1: 1022195-1022195 on Build GRCh38
rs767897458 Chr.1: 1022215-1022215 on Build GRCh38
rs376537367 Chr.1: 1022264-1022264 on Build GRCh38
rs369704878 Chr.1: 1022287-1022287 on Build GRCh38
rs748389976 Chr.1: 1022255-1022255 on Build GRCh38
rs185077738 Chr.1: 1022284-1022284 on Build GRCh38
rs115704555 Chr.1: 1022188-1022188 on Build GRCh38
rs779647850 Chr.1: 1022380-1022380 on Build GRCh38
rs545395744 Chr.1: 1022253-1022253 on Build GRCh38
rs569638601 Chr.1: 1022335-1022335 on Build GRCh38
rs750241246 Chr.1: 1022375-1022375 on Build GRCh38
rs757527582 Chr.1: 1022260-1022262 on Build GRCh38
rs553295392 Chr.1: 1022204-1022204 on Build GRCh38
rs376407731 Chr.1: 1022151-1022151 on Build GRCh38
rs774003955 Chr.1: 1022205-1022205 on Build GRCh38
rs771686237 Chr.1: 1022296-1022296 on Build GRCh38
rs533320704 Chr.1: 1022361-1022361 on Build GRCh38
rs573358968 Chr.1: 1022243-1022243 on Build GRCh38
rs931877780 Chr.1: 1022354-1022354 on Build GRCh38
rs879253787 Chr.1: 1022313-1022313 on Build GRCh38
rs769722871 Chr.1: 1022187-1022187 on Build GRCh38
rs770865981 Chr.1: 1022166-1022166 on Build GRCh38
rs200974261 Chr.1: 1022318-1022318 on Build GRCh38
rs1036873472 Chr.1: 1022184-1022184 on Build GRCh38
rs765443053 Chr.1: 1022357-1022357 on Build GRCh38
rs753197728 Chr.1: 1022295-1022295 on Build GRCh38
rs761387326 Chr.1: 1022214-1022214 on Build GRCh38
rs761473040 Chr.1: 1022285-1022285 on Build GRCh38

View Full Product Details