Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30380501-30380772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPD
Entrez Gene ID
1362
Gene Name
carboxypeptidase D
Gene Aliases
GP180
UniGene
Hs.446079
Gene Chromosome Location
Chr.17: 30378924-30469657 on Build GRCh38
Transcripts
NM_001199775.1 NM_001304.4

SNP Detail

SNP ID Chromosome Location
rs1035244090 Chr.17: 30380607-30380607 on Build GRCh38
rs902785749 Chr.17: 30380549-30380549 on Build GRCh38
rs960874171 Chr.17: 30380617-30380617 on Build GRCh38
rs757168626 Chr.17: 30380531-30380531 on Build GRCh38
rs762250513 Chr.17: 30380659-30380659 on Build GRCh38
rs867467511 Chr.17: 30380645-30380645 on Build GRCh38
rs968692801 Chr.17: 30380732-30380732 on Build GRCh38
rs974130541 Chr.17: 30380693-30380693 on Build GRCh38
rs1025869292 Chr.17: 30380534-30380534 on Build GRCh38
rs778432824 Chr.17: 30380587-30380587 on Build GRCh38
rs537816344 Chr.17: 30380716-30380716 on Build GRCh38
rs745361662 Chr.17: 30380595-30380595 on Build GRCh38
rs1021283081 Chr.17: 30380719-30380720 on Build GRCh38
rs1012384049 Chr.17: 30380623-30380623 on Build GRCh38
rs774862092 Chr.17: 30380608-30380608 on Build GRCh38
rs963111482 Chr.17: 30380646-30380646 on Build GRCh38
rs1019384115 Chr.17: 30380628-30380628 on Build GRCh38

View Full Product Details