Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40695896-40696139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ8B
Entrez Gene ID
79934
Gene Name
coenzyme Q8B
Gene Aliases
ADCK4, NPHS9
UniGene
Hs.130712
Gene Chromosome Location
Chr.19: 40691529-40716885 on Build GRCh38
Transcripts
NM_001142555.2 NM_024876.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5602745 c.1206C>T p.I402I Chr.19: 40695992-40695992 on Build GRCh38
5887475 c.1209+1G>T p.? Chr.19: 40695988-40695988 on Build GRCh38
712843 c.1169G>T p.S390I Chr.19: 40696029-40696029 on Build GRCh38
6370026 c.1188G>T p.E396D Chr.19: 40696010-40696010 on Build GRCh38
7354495 c.1188G>C p.E396D Chr.19: 40696010-40696010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868414197 Chr.19: 40695977-40695977 on Build GRCh38
rs776951694 Chr.19: 40696017-40696017 on Build GRCh38
rs767097657 Chr.19: 40696037-40696037 on Build GRCh38
rs757328390 Chr.19: 40696098-40696098 on Build GRCh38
rs142287240 Chr.19: 40695992-40695992 on Build GRCh38
rs772185208 Chr.19: 40696009-40696009 on Build GRCh38
rs767763269 Chr.19: 40695945-40695945 on Build GRCh38
rs780893463 Chr.19: 40695973-40695973 on Build GRCh38
rs749075139 Chr.19: 40696093-40696093 on Build GRCh38
rs759722843 Chr.19: 40695939-40695939 on Build GRCh38
rs754646727 Chr.19: 40695968-40695968 on Build GRCh38
rs755795213 Chr.19: 40696074-40696074 on Build GRCh38
rs754298223 Chr.19: 40695948-40695948 on Build GRCh38
rs754444229 Chr.19: 40696035-40696035 on Build GRCh38
rs777882752 Chr.19: 40695983-40695983 on Build GRCh38
rs777757961 Chr.19: 40696081-40696081 on Build GRCh38
rs765720942 Chr.19: 40696026-40696026 on Build GRCh38
rs757937743 Chr.19: 40695954-40695954 on Build GRCh38
rs747900552 Chr.19: 40695980-40695980 on Build GRCh38
rs778839711 Chr.19: 40696102-40696102 on Build GRCh38
rs752353148 Chr.19: 40696052-40696052 on Build GRCh38
rs745951936 Chr.19: 40695999-40695999 on Build GRCh38
rs558465263 Chr.19: 40696115-40696115 on Build GRCh38
rs149736065 Chr.19: 40696027-40696027 on Build GRCh38
rs903279907 Chr.19: 40695921-40695921 on Build GRCh38
rs751059382 Chr.19: 40695965-40695965 on Build GRCh38
rs760973154 Chr.19: 40696010-40696010 on Build GRCh38
rs914784859 Chr.19: 40696083-40696083 on Build GRCh38
rs75686781 Chr.19: 40695955-40695955 on Build GRCh38
rs1004573129 Chr.19: 40695935-40695935 on Build GRCh38
rs764405106 Chr.19: 40696016-40696016 on Build GRCh38
rs187371953 Chr.19: 40695947-40695947 on Build GRCh38
rs398122982 Chr.19: 40695998-40695999 on Build GRCh38
rs761030077 Chr.19: 40695946-40695946 on Build GRCh38
rs768670137 Chr.19: 40695992-40695993 on Build GRCh38
rs148712761 Chr.19: 40695989-40695989 on Build GRCh38
rs762358874 Chr.19: 40696018-40696018 on Build GRCh38
rs546628637 Chr.19: 40696113-40696113 on Build GRCh38
rs770968101 Chr.19: 40695991-40695991 on Build GRCh38

View Full Product Details